A better way to meet industrial quality standards in steel particle analysis
Reporting to industrial standards in steel particle analysis is paramount. But traditional optical microscopes provide limited information, and the quality of particle analysis often fails to meet ever-stringent quality requirements.
For a solution, look no further than our Thermo Scientific Phenom ParticleX Steel Desktop SEM. With its automated functions, it empowers you to conduct compositional failure analysis of steel at the nanoparticle level, both during its production, including aluminum-killed, silicon-killed, and calcium-treated processes, and in its finished form such as gear wheels and bearings.
However, what truly differentiates this system is the integrated Thermo Scientific Perception Automation Software. It analyzes polished cross sections of steel samples to automatically find and classify inclusions that can affect the strength, ductility, and corrosion resistance of your steel. Then the Phenom ParticleX Steel Desktop SEM delivers concise industrial reports that turn raw data into clear and unbiased answers for you and your team.
This compact SEM features Thermo Scientific ChemiSEM Technology, our integrated energy-dispersive X-ray spectroscopy (EDS) for elemental research, which enables you to conduct failure analysis and automated characterization of non-metallic inclusions via one simple process.
A better way to meet industrial quality standards in steel particle analysis
Phenom ParticleX Steel Desktop SEM overview
Meet industrial quality standards with confidence
The fast and efficient analysis possible with this system allows you to respond quickly to customer claims of faults and failures. 
Automated unbiased answers
Delivering actionable, ready-to-read reports that summarize the findings in an unbiased way, all with minimal effort needed from the user. 
Efficient and easy-to-use
With a user-friendly interface and integrated automation of routine tasks, this compact desktop system doesn’t just give you faster throughput, it can be mastered quickly by existing and new users alike, with minimal training needed. 
Optimize production with rapid, automated analysis and imaging
The Phenom ParticleX Steel Desktop SEM offers rapid, automated analysis, high-resolution imaging, and detailed compositional data for process adjustments and quality improvements.
Phenom ParticleX Steel Desktop SEM resources
Would you like to know more about our Phenom XL G2 Desktop SEM series? Dive into a wealth of resources, from e-books to datasheets. 
Particle Analysis Learning Center
Our Particle Analysis Learning Center provides comprehensive resources and information on techniques and technologies for analyzing particles in various materials.