1x1 image pixel for data collection
Food Extrusion in Plant-Based Meat Development and Production
Plant-based alternatives to meat
Trust what is inside your meat alternatives
Reverse engineering the characteristics that make animal meats enjoyable to eat using plant-based proteins and other ingredients is no simple feat. You need to be able to mimic a myriad of real meat characteristics—to satisfy not only the senses of sight, smell, and taste—but also provide similar textures, responses to cooking and storage conditions, and nutritional benefits.
On-demand webinar series: Alternative Protein and Plant-based Meat
Watch this webinar series focused on alternative proteins and plant-based meats. 
Plant-based meat ingredients
Meat analogs, created from plant-based ingredients to mimic the fibrous, muscle meat-like texture, taste, appearance, and nutritional value of animal meats, offer a planet-friendly food alternative. Twin-screw extrusion processing is a key technology for continuous production and enhancement of the properties of High Moisture Meat Analog (HMMA) and texturized vegetable protein (texturized proteins) products.
Plant-based meat development
Plant-based meat manufacturing
Reducing the carbon footprint by producing “meat-like“ structures from vegetal proteins to meet the growing demand is great for the planet—reducing carbon emissions and the consumption of animal feed—but it can be a processing challenge. As more consumers not only accept but demand meat analogs, striving to improve the texture and appearance to feel like real meat and achieve a similar eating experience and the mouthfeel is paramount. A twin-screw extrusion process combined with a special die head that can cool the extruded protein in a long flow channel solves that problem and produces a fibrous, plant-based structure similar to real meat to mimic beef, chicken, and seafood.
Extrusion cooking
By applying the precise temperature control and shearing capabilities of a twin-screw extruder, food formulations can be cooked during the process. Extrusion cooking and customized processing parameters enable precisely timed and controlled conditions for protein denaturation, starch gelatinization, flavor encapsulation, and more, all with one technology. With multiple component feeding positions to add recipe ingredients like plant proteins, water, flavors, fats, and micronutrients along with controlled temperature zones for precise heating or cooling, today’s extruders help make alt meat a reality.
3 spoons overflowing with different spices
Twin-screw extrusion process
One technology, two processes
Twin-screw extrusion processing is a key technology for the continuous production of plant-based meat products that mimic animal meats. Two different types of products can be produced with the same twin-screw extrusion technology and slightly different processes:
High moisture meat analogs (HMMA)
Product characteristics: 
Soft and moist with a directly visible fibrous, muscle-meat-like texture.
Process:
 Process set-up for High Moisture Meat Analog (HMMA).
Formation of texture: 
Determined by shear-induced structuring of plant protein formulations in the cooled slit die and the flow characteristics in the die.
Texturized vegetable proteins 
Product characteristics: 
Dry and brittle with a porous structure that rehydrates into a fibrous, meat-like texture.
Process:
Process set-up for texturized vegetable protein.
Formation of texture: 
Determined by pressure drop at the die exit, leading to the evaporation of water and expansion of the product.
Thermo Scientific Twin-Screw Extruders offer a modular solution for application testing and process development from lab- to pilot-scale. All extruder sizes can be equipped with a modular cooled slit die for high moisture meat analogs and a strand-die nozzle for texturized proteins production. These setups promote the creation of a wide range of meat-like textures depending on the combination of formulation, process conditions, and die geometry.
Process 11 Twin-Screw Extruder
During new product development and process optimization, food engineers need highly flexible systems to handle small throughput volumes starting at gram levels up to a few kilograms per hour. They should also be easy to clean, intuitive to operate, and able to mimic a complete processing line. The Thermo Scientific Process 11 Twin-screw Extruder facilitates a streamlined, flexible R&D workflow for a cost-effective, continuous means of production with precise control for maintaining high product quality.
Bench-top twin-screw extruder
Thermo Scientific Process 16 Twin-screw Extruder
A highly modular, flexible extrusion system with a wide range of options and accessories to support your food development, process development, and small batch production requirements. With a wide range of feeding options to accommodate different material types, a stainless steel monocoque housing for easy cleaning, and clearly structured HMI for precise parameter control, the Thermo Scientific Process 16 Extruder allows you to conduct numerous trials efficiently and cost effectively, all while conserving precious laboratory space.
Process 16 Twin Extruder
Food extrusion references
Food extrusion research for developing innovative foods:
These food extrusion publications cover a wide range of research studies related to extrusion technology and its applications in various food products including cereal and snacks, alternative proteins, and biodegradible films to name a few. We are proud that Thermo Scientific extrusion and rheological equipment were used in these studies.  This reaseach aims to further understand the potential of extrusion technology in creating innovative alternative ingredients and foods.  
3 spoons overflowing with different spices
For Research Use Only. Not for use in diagnostic procedures.