Front angle on the ProFlex PCR System, including the user interface home screen
The ProFlex PCR System combines the reliability and performance you’ve come to expect from Applied Biosystems thermal cyclers with the flexible configuration and control features that fit how you work today. Interchangeable block formats allow you to maximize your throughput or run independent experiments concurrently. And now you can access your ProFlex PCR System with Thermo Fisher Connect, giving you the freedom to design and securely upload your methods and monitor runs from any smartphone or desktop computer.
Meet the next-generation PowerFlex thermal cycler
Engineered to deliver exceptional performance, compatibility, and productivity for PCR workflows of any applications.
Discover more
ProFlex thermal cyclers to maximize your throughput with flexibility
Maximize your throughput using Proflex PCR system that supports up to 5 interchangeable blocks based on your needs.
PCR system with interchangeable blocks for high throughput PCR or simultaneous experiments
The ProFlex PCR System has five different blocks that can be changed with the flip of a switch, including the first-of-its-kind 3 x 32-well block. This block allows up to three experiments to be run simultaneously, completely independently of each other and without interference. Dual 96-well and dual 384-well blocks are available for your high-throughput PCR sequencing needs, and a dual flat block is available to support Applied Biosystems OpenArray plate technology for genotyping analysis on the Applied Biosystems QuantStudio 12K Flex Real-Time PCR System.
Top of the ProFlex PCR System containing the 3 x 32-well block
ProFlex PCR System with the 3 x 32-well block for running up to three independent experiments simultaneously.
Top and partial screen of the ProFlex PCR System containing the Dual 384-well block
Dual 384-well block for higher-throughput sequencing using ProFlex PCR Systems.
Top of the ProFlex PCR System containing the Dual flat block
Dual flat block on the ProFlex PCR System supports OpenArray plate technology for genotyping analysis on the QuantStudio 12K Flex Real-Time PCR System.
Top and screen of the ProFlex PCR System containing the 96-well block
96-well block for standard PCR applications using the ProFlex thermal cycler.
Top of the ProFlex PCR System containing the Dual 96-well block
Dual 96-well block for the ProFlex thermal cycler for high-throughput sequencing.
How to unbox and set up ProFlex thermal cyclers
Ordering information
Resources
Thermal Cycler Software and Firmware Files
Download the latest updates for your instrument
Interested in private label thermal cyclers?
Submit an inquiry
Support
Service
We provide a full range of service options for all Applied Biosystems thermal cyclers.
View service plans
Multiplex Dynamic Temperature Verification Service
Instrument qualification service: Have one of our engineers perform temperature qualifications at your site.
View temperature service options
 Download MDTV flyer
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments