Embrace the future and explore new frontiers of scientific discovery right from your lab bench. With a comprehensive collection of QuantStudio Real-Time PCR (qPCR) systems to choose from, we empower you to shape the future of your research according to your own strategies and budget. Our continuous innovation in qPCR technology yields ongoing advancements to maximize performance across the QuantStudio portfolio, including:
  • Faster results
  • Smarter workflows
  • Exceptional data quality
  • High efficiency
Save now with no interest financing on instruments
Capabilities
Precise quantification, with 1.5-fold discrimination
Uncover a richer data set with 1.5-fold discrimination. Data shows distinct clusters indicating 2-fold and 1.5-fold discrimination on QuantStudio 7 Pro system with 99.9% confidence, equivalent to other Applied Biosystems platforms.
...
Intuitive touch screens
From easy set-up to monitoring reactions in real-time. QuantStudio systems enable visualization of your RT-PCR reaction in real-time. The graphical interface simplifies use and provides easy to understand status. 
...
Robust, friendly software
Design and Analysis 2 Software is designed to create and edit the qPCR run setup for multiple instrument types. It offers single-plate data analysis for CT, standard curve, relative quantification, genotyping and presence/absence of gene of interest.
...
Run multiple optimizations simultaneously
Most QuantStudio systems with 96-well block format feature VeriFlex technology, which provides precise independent temperature control in each zone. Blocks contain either three or six VeriFlex zones. Each zone can be a different temperature, allowing multiple temperature optimizations in the same run.
...
Access, analyze, and share data anytime, anywhere
Remotely monitor your runs, quickly analyze sophisticated data sets, store results in a secure space, and safely share them online with colleagues across campus and around the world with Web browser-based software supported by our Connect cloud-based platform.
...
Hear what our customers have to say
"The interaction with instruments through voice command is highly appreciated because typically the operator has something in their hands or gloves and so forth.” 
—Mikael Kubista, Head and Founder of TATAA Biocenter AB, Sweden
...
Looking for commercial-use products & services?