QuEChERS sample preparation—now easier, more convenient, and affordable
QuEChERS (Quick, Easy, Cheap, Effective, Rugged, and Safe) is a sample preparation method that was initially developed in 2003 for analyzing veterinary drugs in food. This innovative method was also successfully tested for pesticide residue analysis in plant materials, demonstrating its versatility and effectiveness. The detailed method was published in the journal AOAC International in 2003. Since then, the QuEChERS method has evolved into a widely adopted sample preparation technique used for a variety of analytes across an expanding number of sample types before analysis by gas chromatography-mass spectrometry (GC-MS) or liquid chromatography-mass spectrometry (LC-MS).
Thermo Scientific QuEChERS kits are designed to streamline your workflow and minimize the potential for error by providing pre-packaged, ready-weighed salts, sorbents, and buffers. These kits are tailored to your specific analytes of interest, sample matrix, and preferred QuEChERS method, ensuring optimal performance and convenience. The available methods include:
✓Original 2003 Method: The foundational method that started it all, providing a robust and reliable approach to sample preparation.
✓AOAC 2007.01 Method: A refined version of the original method, offering enhanced performance for specific analytes and matrices.
✓EN 15662 Method: A European standard method that provides a comprehensive approach to pesticide residue analysis.
✓NY/T 1380-2007 Method: A method tailored to specific regulatory requirements and analytical needs.
Thermo Scientific offers complete packages that include both extraction and clean-up kits, as well as extraction-only and clean-up-only kits to suit your laboratory’s specific sample preparation needs. These pre-packaged kits save time and money, simplify QuEChERS procedures, and ensure high-quality results. By using these kits, you can achieve reproducible results and excellent recoveries for a wide variety of analytes, making your analytical testing more efficient and reliable.
As shown in the workflow diagram below, the QuEChERS method comprises four basic steps:
  1. Homogenize Sample (Fruit or Vegetable): Begin by thoroughly homogenizing the sample, such as a fruit or vegetable, to ensure a uniform consistency. This step is crucial for accurate and representative analysis, as it ensures that the analytes are evenly distributed throughout the sample.
  2. Extract Analytes of Interest: Extract the analytes of interest from the homogenized sample using specific extraction salts. These analytes can include pesticide residues, veterinary drugs, and/or mycotoxins. The extraction process involves adding the salts to the sample, which facilitates the release of the target compounds into the solution.
  3. Clean-Up the Resulting Supernatant: Clean up the resulting supernatant using dispersive solid phase extraction (dSPE). This step involves adding sorbents to the supernatant to remove unwanted matrix components that could interfere with the analysis. The dSPE process helps to purify the extract, ensuring that the final sample is free from contaminants and ready for accurate analysis.
  4. Analyze by GC-MS or LC-MS: Finally, analyze the cleaned-up extract using gas chromatography-mass spectrometry (GC-MS) or liquid chromatography-mass spectrometry (LC-MS). These powerful analytical techniques allow for the precise identification and quantification of the target analytes. The choice between GC-MS and LC-MS depends on the specific properties of the analytes and the requirements of the analysis.
By following these four basic steps, the QuEChERS method provides a streamlined and efficient approach to sample preparation, enabling reliable and reproducible results for a wide range of analytes.
Typical QuEChERS Workflow
A Range of QuEChERS extraction kits for original 2003 method
The original QuEChERS method (Anastassiades et al., 2003) was developed by Steven J. Lehotay and Michelangelo Anastassiades in 2003 for the determination of pesticide residues in fruits and vegetables. This pioneering method was specifically designed to analyze non-base-sensitive compounds and employs a sodium chloride buffer to reduce polar interferences and enhance the efficiency of the extraction process.
Buffering is a critical component of the QuEChERS procedure because it allows for the accurate analysis of pesticides and other residues that display pH dependence in their recoveries, depending on the sample matrix. The buffering process helps to stabilize the pH levels, ensuring that the target analytes are effectively extracted and accurately quantified.
To support the original 2003 QuEChERS method, three different extraction kits are available
Original 2003 method extraction kits
 Part numberExtraction saltsAlso includesStarting sampleUnit sizePurchase
S1-10-ORIG-CH-KITS1-10-ORIG-CH-KIT
  • 4 g MgSO4
  • 1 g NaCl
  • In pouches
  • 50 mL tubes
  • Ceramic homogenizers
10 g sample50/PK
Buy now
S1-10-ORIG-KITS1-10-ORIG-KIT
  • 4 g MgSO4
  • 1 g NaCl
  • In pouches
50 mL tubes10 g sample50/PK
Buy now
S1-10-ORIG-POTS1-10-ORIG-POT
  • 4 g MgSO4
  • 1 g NaCl
  • In pouches
̶10 g sample50/PK
Buy now
S1-15-ORIG-CH-KITS1-15-ORIG-CH-KIT
  • 6 g MgSO4
  • 1.5 g NaCl
  • In pouches
  • 50 mL tubes
  • Ceramic homogenizers
15 g sample50/PK
Buy now
S1-15-ORIG-KITS1-15-ORIG-KIT
  • 6 g MgSO4
  • 1.5 g NaCl
  • In pouches
50 mL tubes15 g sample50/PK
Buy now
S1-15-ORIG-POTS1-15-ORIG-POT
  • 6 g MgSO4
  • 1.5 g NaCl
  • In pouches
-15 g sample50/PK
S1-10-ORIG-KIT
Buy now
Chemicals used in the original 2003 method
Acetonitrile:Used to perform single-phase extraction (SPE) on a sample. Acetonitrile is a polar organic solvent that effectively extracts a wide range of analytes, including pesticides, veterinary drugs, and mycotoxins, from the sample matrix. Its miscibility with water makes it ideal for extracting both polar and non-polar compounds, ensuring comprehensive analyte recovery.
Anhydrous MgSO4:Induces phase separation between the water content in the sample and the acetonitrile layer during the liquid-liquid partitioning step. This separation is crucial for isolating the analytes in the organic phase.
Removes remaining water from the sample during the dispersive solid phase extraction (dSPE) step. Anhydrous magnesium sulfate acts as a drying agent, ensuring that the final extract is free from excess moisture, which could interfere with subsequent analytical steps.
NaCl:Induces phase separation between the water content in the sample and the acetonitrile layer during the liquid-liquid partitioning step. Sodium chloride helps to increase the ionic strength of the aqueous phase, promoting the partitioning of analytes into the acetonitrile layer.
Removes residual water from the sample, enhancing the efficiency of the extraction process.
Reduces polar interference and enhances extraction by minimizing the solubility of polar compounds in the aqueous phase, thereby improving the recovery of target analytes in the organic phase.
PSA (Primary Secondary Amine):Used to remove polar matrix components, citric co-extractives, and free fatty acids from the sample during the dispersive solid phase extraction (dSPE) step. PSA is a sorbent that selectively binds to and removes these interfering substances, ensuring that the final extract is clean and suitable for accurate analysis by GC-MS or LC-MS.
QuEChERS extraction and clean-up kits for AOAC 2007.01 method
The AOAC 2007.01 QuEChERS Method, developed by Steven J. Lehotay and his co-workers, refines the original 2003 method by incorporating higher concentrations of acetate buffering for enhanced buffering strength. Buffering is essential in the QuEChERS procedure as it stabilizes pH during extraction and clean-up steps, ensuring accurate analysis of pH-dependent pesticides and other residues.
The method starts with homogenizing the sample, such as fruits or vegetables, to achieve uniform consistency. Extraction salts, including magnesium sulfate and sodium acetate, are then added to release analytes into the acetonitrile solvent. The mixture undergoes liquid-liquid partitioning, where anhydrous magnesium sulfate and sodium chloride induce phase separation and remove residual water.
The supernatant is cleaned using dispersive solid phase extraction (dSPE) with sorbents like primary secondary amine (PSA) to remove polar matrix components. The purified extract is then analyzed by GC-MS or LC-MS for precise identification and quantification. Thermo Scientific provides pre-packaged kits to support this method, streamlining workflows and ensuring high-quality, reproducible results.
AOAC 2007.01 method extraction kits
 Part numberExtraction saltsAlso includesStarting sampleUnit sizePurchase
S1-15-AOAC-CH-KITS1-15-AOAC-CH-KIT
  • 6 g MgSO4
  • 1.5 g NaOAc
  • In pouches
  • 50 mL tubes
  • Ceramic homogenizers
  • 15 g sample
  • 50/PK
Buy now
S1-15-AOAC-KIT
S1-15-AOAC-KIT
  • 6 g MgSO4
  • 1.5 g NaOAc
  • In pouches
  • 50 mL tubes
  • 15 g sample
  • 50/PK
Buy now
S1-15-AOAC-POT
S1-15-AOAC-POT
  • 6 g MgSO4
  • 1.5 g NaOAc
  • In pouches
̶
  • 15 g sample
  • 50/PK
Buy now
AOAC 2007.01 method clean-up kits
 Sample typePrefilled 2 mL tubes
(100/PK)
Prefilled 15 mL tubes
(50/PK)
  AOAC 2007.01 method clean-up kits
Clean-up
General fruits and vegetablesS2-2-GFV-AOAC-KIT
  • 50 mg PSA
  • 150 mg Anhydrous MgSO4
S2-15-GFV-AOAC-KIT
  • 400 mg PSA
  • 1200 mg Anhydrous MgSO4
Pigmented fruits and vegetablesS2-2-P-AOAC-KIT
  • 50 mg PSA
  • 50 mg GCB
  • 150 mg MgSO4
S2-15-P-AOAC-KIT
  • 400 mg PSA
  • 400 mg C18
  • 1200 mg Anhydrous MgSO4
Fruits and vegetables with fats and waxesS2-2-FW-AOAC-KIT
  • 50 mg PSA
  • 50 mg C18
  • 150 mg Anhydrous MgSO4
S2-15-FW-AOAC-KIT
  • 400 mg PSA
  • 400 mg C18
  • 1200 mg Anhydrous MgSO4
Fruits and vegetables with pigments and fatsS2-2-PF-AOAC-KIT
  • 50 mg PSA
  • 50 mg C18
  • 50 mg GCB
  • 150 mg Anhydrous MgSO4
S2-15-PF-AOAC-KIT
  • 400 mg PSA
  • 400 mg C18
  • 400 mg GCB
  • 1200 mg Anhydrous MgSO4
All food typesS2-2-ALL-AOAC-KIT
  • 50 mg PSA
  • 50 mg C18
  • 7.5 mg GCB
  • 150 mg MgSO4
S2-15-ALL-AOAC-KIT
  • 400 mg PSA
  • 400 mg C18
  • 45 mg GCB
  • 1200 mg MgSO4
Other foodsS2-2-OTH-AOAC-KIT
  • 25 mg C18
  • 150 mg MgSO4
S2-15-OTH-AOAC-KIT
  • 150 mg C18
  • 900 mg MgSO4
Chemicals used in the AOAC 2007.01 method
Acetonitrile:Used to perform single-phase extraction on a sample. It effectively extracts a wide range of analytes, including pesticides, veterinary drugs, and mycotoxins. Its miscibility with water allows for efficient extraction of both polar and non-polar compounds.
Anhydrous MgSO4:Induces phase separation between the water content in the sample and the acetonitrile layer during the liquid-liquid partitioning step. It removes remaining water from the sample during the dispersive solid phase extraction (dSPE) step, acting as a drying agent to ensure the final extract is free from excess moisture.
NaOAc (sodium acetate):Buffers the sample to stabilize pH. This buffering is essential for maintaining consistent pH levels during the extraction process, ensuring accurate recovery of pH-dependent analytes.
PSA (primary secondary amine):
Used during the dSPE step to remove polar matrix components, citric co-extractives, and free fatty acids from the sample, ensuring that the final extract is clean and suitable for accurate analysis.
C18:Removes fats, sterols, and other non-polar interferences from the sample, reducing the presence of non-polar contaminants that could interfere with the analysis.
GCB (graphitized carbon black):Removes pigments from the sample. It is not recommended for use with planar pesticides as it may also remove these target analytes. GCB is particularly effective in cleaning up samples with high pigment content, ensuring a clearer extract for analysis.
QuEChERS extraction and clean-up kits for EN 15662 method
The extraction and clean-up products here support the EN 15662 method. Widely used in Europe, the EN 15662 method was developed by Michelangelo Anastassiades and co-workers, who modified the original 2003 method to use citrate buffering at a relatively low buffering capacity. This variation employs a sodium chloride buffer to minimize polar interferences, along with sodium citrate dihydrate and disodium citrate sesquihydrate buffers. Buffering is a crucial part of the QuEChERS procedure because it stabilizes the pH, allowing for accurate analysis of pesticides and other residues that exhibit pH dependence in their recoveries, depending on the sample matrix. This ensures consistent and reliable results across various sample types.
EN 15662 method extraction kits
 Part numberExtraction saltsAlso includesStarting sampleUnit sizePurchase
S1-10-EN-CH-KITS1-10-EN-CH-KIT
  • 4 g MgSO4
  • 1 g NaCl
  • 0.5 g disodium hydrogencitrate sesquihydrate
  • 1 g trisodium citrate dihydrate
  • In pouches
  • 50 mL tubes
  • Ceramic homogenizers
10 g50/PK
Buy now
S1-10-EN-KITS1-10-EN-KIT
  • 4 g MgSO4
  • 1 g NaCl
  • 0.5 g disodium hydrogencitrate sesquihydrate
  • 1 g trisodium citrate dihydrate
  • In pouches
50 mL tubes10 g50/PK
Buy now
S1-10-EN-POTS1-10-EN-POT
  • 4 g MgSO4
  • 1 g NaCl
  • 0.5 g disodium hydrogencitrate sesquihydrate
  • 1 g trisodium citrate dihydrate
  • In pouches
-10 g50/PK
Buy now
EN 15662 method clean-up kits
 Sample typePrefilled 2 mL tubes
(100/PK)
Prefilled 15 mL tubes
(50/PK)
EN 15662 method clean-up kit
Clean-up
General fruits and vegetables
S2-2-GFV-EN-KIT
  • 150 mg MgSO4
  • 25 mg PSA
S2-15-GFV-EN-KIT
  • 900 mg MgSO4
  • 150 mg PSA
Pigmented fruits and vegetablesS2-2-P-EN-KIT
  • 150 mg MgSO4
  • 25 mg PSA
  • 2.5 mg GCB
S2-15-P-EN-KIT
  • 900 mg MgSO4
  • 150 mg PSA
  • 15 mg GCB
Highly pigmented fruits and vegetables
S2-2-HP-EN-KIT
  • 150 mg MgSO4
  • 25 mg PSA
  • 7.5 mg GCB
S2-15-HP-EN-KIT
  • 900 mg MgSO4
  • 150 mg PSA
  • 45 mg GCB
Fruits and vegetables with fats and waxes
S2-2-FW-EN-KIT
  • 150 mg MgSO4
  • 25 mg PSA
  • 25 mg C18
S2-15-FW-EN-KIT
  • 900 mg MgSO4
  • 150 mg PSA
  • 150 mg C18
Chemicals used in the EN 15662 method
Acetonitrile:Used to perform single-phase extraction on a sample. It serves as a polar organic solvent that effectively extracts a wide range of analytes, including pesticides and residues, from the sample matrix. Its miscibility with water allows for efficient extraction of both polar and non-polar compounds.  
Anhydrous MgSO4:Induces phase separation between the water content in the sample and the acetonitrile layer during the liquid-liquid partitioning step. Additionally, anhydrous magnesium sulfate removes remaining water from the sample during the dispersive solid phase extraction (dSPE) step, acting as a drying agent to ensure the final extract is free from excess moisture.
NaCl:Induces phase separation between the water content in the sample and the acetonitrile layer during the liquid-liquid partitioning step and helps to remove residual water from the sample. This step enhances the efficiency of the extraction process by reducing polar interference
PSA (Primary Secondary Amine):Used during the dSPE step to remove polar matrix components, citric co-extractives, and free fatty acids from the sample, ensuring that the final extract is clean and suitable for accurate analysis.
C18:Used to remove fats, sterols, and other non-polar interferences from the sample, reducing the presence of non-polar contaminants that could interfere with the analysis.
GCB (Graphitized Carbon Black):Used to remove pigments from the sample. However, it is not recommended for use with planar pesticides as it may also remove these target analytes. GCB is particularly effective in cleaning up samples with high pigment content, ensuring a clearer extract for analysis.
QuEChERS clean-up kit for NY/T 1380-2007 method
The QuEChERS clean-up kit for the NY/T 1380-2007 method is recommended by the China Ministry of Agriculture. This method is designed for the sample preparation of 51 specific pesticide residues in fruits and vegetables, ensuring accurate and efficient analysis by GC-MS. It streamlines the process for reliable results.
NY/T 1380-2007 method clean-up kit
 SamplePrefilled 15 mL tubes
(50/PK)
Purchase
NY/T 1380-2007 method clean-up kitFruits and vegetablesS2-15-GFV-NYT-KIT
  • 100 mg PSA
  • 100 mg C18
  • 300 mg Anhydrous MgSO4
Buy now
Chemicals used in the NY/T 1380-2007 method
Anhydrous MgSO4:Induces phase separation between the water content in the sample and the acetonitrile layer during the liquid-liquid partitioning step. It also removes remaining water from the sample during dispersive solid phase extraction (dSPE).  
PSA (Primary Secondary Amine):Used during the dSPE step to remove polar matrix components, citric co-extractives, and free fatty acids from the sample, ensuring a clean extract for accurate analysis.
C18:Employed to remove fats, sterols, and other non-polar interferences from the sample, reducing the presence of contaminants that could interfere with the analytical results.
Part numberProduct description
Vials & caps
6ASV9-1P2mL clear screw glass vial (9mm short thread) with write-on patch
6ESV9-1PP1.5mL clear screw PP vial (9mm short thread)
6ASV9-2P2mL amber screw glass vial (9mm short thread) with write-on patch
6ASC9ST1RRed screw cap (9mm) (PP: white silicone/red PTFE septa: 1.0mm thickness) with AVCS technology
6ASC9ST1Blue screw cap (9 mm) (PP: white silicone/blue PTFE septa: 1.0 mm thickness) with AVCS technology
Part numberProduct description
Solvents
A956-1Thermo Scientific UHPLC-MS grade acetonitrile suitable for LC and GC
A458-1Thermo Scientific UHPLC-MS grade methanol suitable for LC and GC
W8-1Thermo Scientific UHPLC-MS grade water suitable for LC and GC
A117Fisher Chemical Optima LC-MS grade formic acid suitable for LC and GC
A115-50Fisher Chemical Optima LC-MS grade ammonium formate suitable for LC and GC
T00101400Thermo Scientific Chromplete acetonitrile suitable for HPLC and GC
D1514Optima Methylene Chloride for suitable HPLC and GC-MS
H3064Optima 95% n-Hexane suitable for HPLC and GC-MS
S415500Sodium sulfate anhydrous suitable for LC and GC
Part numberProduct description
HPLC columns
17326-102130Thermo Scientific Accucore aQ (100 × 2.1 mm, 2.6 μm)
071387Thermo Scientific Acclaim Trinity P1 (anionic pesticides) (100 × 3.0 mm, 3.0 µm)
17626-102130Thermo Scientific Accucore RP-MS (100 × 2.1 mm, 2.6 μm)
17326-102130Thermo Scientific Accucore aQ (100 × 2.1 mm, 2.6 μm)
Part numberProduct description
GC consumables
453T2120Thermo Scientific LinerGold: Siltek six baffle PTV liner
453A1925UISSL splitless liner, single taper, deactivated
26096-1420Thermo Scientific TraceGOLD TG-5SIL MS (30 m x 0.25 mm i.d. x 0.25 μm)
Style Sheet for Komodo Tabs
Style Sheet for Global Design System
CMD SchemaApp code