Nicolet RaptIR+/RaptIR FTIR Microscopes
In analytical labs where details matter most—when productivity is measured in microns and minutes—turn to the Thermo Scientific Nicolet RaptIR or RaptIR+ FTIR Microscope for detailed answers.
These research-grade microscopes help you rapidly locate and identify trace materials, inclusions, impurities, microparticles, and determine the distribution of materials within a sample with incredible clarity and precision. Its clean, adaptive user interface provides increased flexibility and usability for all users, from specialists to novices. The Nicolet RaptIR and RaptIR+ FTIR Microscopes provide answers quickly, regardless of industry or application.
Expanding your options, the RaptIR+ adds the versatility of user-swappable detectors offering extended-range operation or liquid-nitrogen-free operation. When adapted to interface with a Nicolet iS50 FTIR Spectrometer, the RaptIR+ enables a wide range of new analysis possibilities, including TGA-IR, GC-IR, NIR, FT-Raman, and much more. For a more cost-conscious choice, the RaptIR+ can also adapt to the Nicolet Apex FTIR Spectrometer.
Features
From the mosaic feature to infrared capability and a seamless user experience, the Nicolet RaptIR and RaptIR+ FTIR Microscopes are full of technological innovations that help specialists and non-specialists alike.
  • Users can find and analyze the smallest samples in as little as 90 seconds.
  • Diffraction-limited objectives provide outstanding visual and infrared performance, generating high-quality, high-resolution spectra of sample with visual spatial resolution of 1 micron and infrared spatial resolution of 5 microns.
  • A multi-position automated nosepiece, autofocus, auto-illumination, and full-view computer-controlled aperture combine to deliver an easy-to-use, efficient and comfortable user experience.
  • Reflectance, transmission, and ATR sampling modes with automated exchange provide versatility, while a built-in ATR pressure sensor automatically indicates when there is ATR contact.
  • An open-access, heavy-duty stage accommodates samples up to 40 millimeters thick and 5 kilograms in weight.
  • Mapping at a rate of up to 10 spectra/second provides results quickly.
  • The Nicolet RaptIR+ adds even more exciting features, including detector choices and configurations permitting the maximum useability of an attached spectrometer.
Video Player is loading.
Current Time 0:00
Duration 6:53
Loaded: 0.00%
Stream Type LIVE
Remaining Time 6:53
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Nicolet RaptIR+ FTIR Microscope Walkthrough
    FTIR spectrometer combined with FTIR microscope
    Nicolet RaptIR+ with iS50 FTIR Spectrometer
    The Nicolet RaptIR and RaptIR+ FTIR Microscopes are always coupled to a Thermo Scientific Nicolet FTIR Spectrometer. Using it with the Thermo Scientific Nicolet iS50 FTIR Spectrometer provides a broader array of capabilities—including FT-Raman, TGA-IR, NIR, far-infrared, and diamond ATR—for addressing complex analytical challenges. This combination will help you find answers more quickly, whether you are studying powders, liquids, gases, or micron-sized particulates.
    Find the best Nicolet RaptIR FTIR Microscope to fit your needs with this comparison guide
     Nicolet RaptIR+ FTIR MicroscopeNicolet RaptIR FTIR Microscope
    Visible resolution
    1 micron1 micron
    Infrared resolution
    5 micron5 micron
    Multiple detectors
    Swappable, MCT-A, MCT-B, InGaAsFixed, MCT-A, MCT-B
    Spectrometer option
    Nicolet iS50 FTIR Spectrometer (right or left side)
    Nicolet Apex FTIR Spectrometer (right side)
    Nicolet iS50 FTIR Spectrometer (right side)
    Objectives
    4X or 40X visible, 15X or grazing angle infrared4X or 40X visible, 15X or grazing angle infrared
    Polarizers
    Motorized, visible and IRMotorized, visible and IR
    The Nicolet RaptIR FTIR Microscope's adaptable design is suitable for scientists across all industries and skill levels. It has been engineered with the latest technology to ensure you get sharp images and rapid, accurate analysis at every step of your workflow.
    The Nicolet RaptIR+ FTIR Microscope automatically ensures you can use it anywhere, anytime.
    Load samples up to 40 millimeters thick and 5 kilograms in mass with the open access and heavy-duty stage.
    The high-resolution camera and eyepieces display clear images, and the adaptive UI and comfortable joystick provide a nearly effortless user experience.
    The 4x and 15x objectives enable you to get a clear view of your entire sample and focus in on the smallest detail.
    Cover a large area faster than ever with the microscope's infrared capability and examine your sample with up to 10 spectra per second at a resolution greater than 5 microns.
    Paradigm Software 64-bit operating system and UI simplify your entire analytical process.
    The microscope facilitates the transmission of data into a wide variety of formal and fully customizable formats to send to your stakeholders.
    The Nicolet RaptIR+ FTIR Microscope automatically ensures you can use it anywhere, anytime.
    Load samples up to 40 millimeters thick and 5 kilograms in mass with the open access and heavy-duty stage.
    The high-resolution camera and eyepieces display clear images, and the adaptive UI and comfortable joystick provide a nearly effortless user experience.
    The 4x and 15x objectives enable you to get a clear view of your entire sample and focus in on the smallest detail.
    Cover a large area faster than ever with the microscope's infrared capability and examine your sample with up to 10 spectra per second at a resolution greater than 5 microns.
    Paradigm Software 64-bit operating system and UI simplify your entire analytical process.
    The microscope facilitates the transmission of data into a wide variety of formal and fully customizable formats to send to your stakeholders.
    The versatility of the RaptIR+/RaptIR allows you to analyze across a wide range of applications:
    • Basic analysis—Work to identify contamination in production materials and find the origin of embedded particles.
    • Pharmaceuticals—Collect high-quality IR data over an entire tablet to validate distribution/layering of a time-release dosage.
    • Environment—Examine a dense microplastic sample with a wide field of view, then target a specific particle with automated visible and IR features.
    • Art restoration—Validate the source of materials in a painting or ancient text to gain insights into provenance and plan preservation activities.
    • Forensics—Work to identify paints, inks, dyes, biomaterials in cloth, fibers, or paper to provide evidence of fraud, examine suspected documents, or link trace evidence from various sources.
    Thermo Scientific Nicolet RaptIR FTIR Microscope Product Tour
    See it fast. Identify it faster. Watch this video to experience the Nicolet RaptIR FTIR Microscope—designed with enhanced precision and agility to help streamline analysis and generate actionable results with amazing speed.
    Video Player is loading.
    Current Time 0:00
    Duration 2:09
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 2:09
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Thermo Scientific OMNIC Paradigm Software allows you to swiftly navigate and analyze complex samples. It employs a database format for collecting data, utilizes the full power of the Windows 10 operating system, and collects impressively large maps to enable you to collect multiple regions of interest.
      The innovative hardware features of the Nicolet RaptIR+ FTIR Microscope are complemented by the advantages of the OMNIC Paradigm Desktop Software. It provides users with clear guidance on how to extract information needed from the target.
      The software boasts a clean, adaptive user interface (UI), database security, floating tool bar, 64-bit system, and easily generated workflows to automate data collection and analysis sequences.
      Learn more about OMNIC Paradigm Software >
      See samples with stunning clarity
      10 micron beads embedded in an epoxy
      10 micron beads embedded in an epoxy
      Fibers finely resolved and analyzed
      Fibers finely resolved and analyzed
      Incredible detail of ink on a plastic surface
      Incredible detail of ink on a plastic surface
      Advantages in pharmaceutical analysis
      Examine entire tablets
      Examine entire tablets
      Gain insight about API distribution
      Gain insight about API distribution
      Performance in analytical labs
      Colors of complex images
      Colors of complex images
      Forensic sample details - auto part contamination
      Forensic sample details - auto part contamination
      Forensic sample details - paint chip analysis
      Forensic sample details - paint chip analysis
      Microplastics analytical capabilities
      Analyze the environmental impact of microplastics
      Analyze the environmental impact of microplastics
      The software’s clean, adaptive user interface allows specialists and novice users alike to obtain the data they’re looking for. Workflow automation, combined with the ability to select multiple regions of interest in a sample for collection in one pass, helps save time and reduce errors. Autofocus and automated illumination allow for quick and easy data collection. Fully customizable microparticles analysis workflow allows users of varying experience to click, get images, and move forward, choosing what they want to show in tailored reports.
      See for yourself in these short videos how easy it is to use the Nicolet RaptIR+/RaptIR FTIR Microscope to find a sample, collect a mosaic, or collect a background and map:
      Video Player is loading.
      Current Time 0:00
      Duration 0:55
      Loaded: 0.00%
      Stream Type LIVE
      Remaining Time 0:55
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Find your sample fast using software
        Video Player is loading.
        Current Time 0:00
        Duration 0:54
        Loaded: 0.00%
        Stream Type LIVE
        Remaining Time 0:54
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          Collect the background and an image map of your sample
          Video Player is loading.
          Current Time 0:00
          Duration 0:38
          Loaded: 0.00%
          Stream Type LIVE
          Remaining Time 0:38
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
            • en (Main), selected
            Collect a high-resolution mosaic using software
            Video Player is loading.
            Current Time 0:00
            Duration 1:52
            Loaded: 0%
            Stream Type LIVE
            Remaining Time 1:52
            1x
              • Chapters
              • descriptions off, selected
              • captions off, selected
              • en (Main), selected
              Analysis of a high-res multi-layer film
              Nicolet RaptIR and RaptIR+ resources
              Microspectroscopy application, smart, and tech notes:
              Contact us today to learn how you too can obtain answers in minutes and microns with the Nicolet RaptIR+/RaptIR FTIR Microscopes and Thermo Scientific OMNIC Paradigm Software.
              Please complete the form below and we will contact you shortly.
              * Required field
               I want to consult with an expert.
               I'd like a product quote.
               I'd like information on Installation and Training Services.
              *
              *
              *
              *
              *
              *
              *
               Additive Manufacturing
               Art and Antiquities
               Biomaterials
               Ceramics/Glass/Displays
               Chemicals/Petrochemicals
               Composite Materials
               Cosmetics/Personal Care Products
               Energy-related Materials
               Foods/Beverages
               Forensic Evidence/Reference Samples
               Gem and Geological Samples
               Metals/Alloys
               Microplastics
               Minerals/Cement
               Nanomaterials
               Nucleic Acid
               Pharmaceuticals/Biopharma/Nutraceuticals
               Polymers/Plastics
               Proteins
               Semiconductors/Electronics
               None of these are applicable to me
               I would like to receive marketing and promotional emails regarding products and services from Thermo Fisher Scientific.
              This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
              Style Sheet for Global Design System