SpeedVac family image
Pursue new discoveries with speed and precision
Samples are precious and so is time. Thermo Scientific Savant SpeedVac vacuum concentrators, refrigerated vapor traps and pumps use state-of-the-art centrifugation, vacuum and heat technologies for removing solvents and concentrating samples while maintaining sample integrity. Choose from any of the nine different models of vacuum concentrator to suit your needs from biological to non-biological samples from low to high volume samples.
Featured SpeedVac Vacuum Concentrator Categories
SpeedVac SRF110 modular refrigerated vacuum concentrator provides flexibility to benchtop space. It offers unique programming capability to maintain chamber temperature pre- and post-sample run to prevent thermal degradation of heat sensitive samples such as proteins, enzymes, and other biomolecules.
From traditional drying down of DNA preparations in water and methanol, to the more complex and aggressive applications presented by the combinatorial chemistry and drug discovery sectors of the pharmaceutical industry, we have the systems for your lab. 
Individual vacuum concentrators for upgrading an existing piece of equipment or when you are looking to build your system from scratch.
Refrigerated Vapor Traps are available with two temperature ranges from ambient temperature to -50°C and  ambient temperature to -105°C
Savant Universal Vacuum Systems are the most energy efficient and environmentally safe vacuum system available, providing complete recovery of evaporated solvents
Rotors are available in a wide variety of styles and sizes to accommodate all your sample requirements
Two pump styles are available from Thermo Scientific: traditional rotary vane oil pumps and the newer oil-free, maintenance free vacuum pumps
Chemical traps are highly recommended to absorb residual volatile vapors that may bypass the primary cold trap
Improve the functionality of Thermo Scientific Savant SpeedVac Concentrators with Savant SpeedVac concentrator accessories
Why use centrifugation for evaporation?
Centrifugation generates a centrifugal force to prevent bumping (bubbling), boiling, and physical loss of the sample. To completely dry the sample, the solute is deposited at the bottom of the container for complete quantitative recovery.
SpeedVac solvent evaporation examples
group-1-Blue-Water-flat-175
SPD140DDA, VLP120, RVT400 4ml Blue H2O, 65°C, no lights after 1 hour
group-2-Blue-Water-flat-175
SPD140DDA, VLP120, RVT400 4ml, Blue H2O, 65°C, no lights Vac 14 torr after 1 hour
group-3-Blue-Water-flat-175
SPD2030 4ml, Blue H2O, 65°C, no lights Vac 0.1 torr after 1 hour
group-4-Blue-Water-flat-175
SPD140DDA, VLP120, RVT400 4ml, Blue H2O, 65°C, no lights Vac 0.1 torr after 2 hours
Vacuum promotes solvent evaporation in the SpeedVac chamber. Samples are maintained in liquid state at sub-ambient temperature throughout the concentration process, preventing loss of activity or damage to heat sensitive substances. By applying a controlled vacuum the boiling point of the solvents can be lowered to the point that the liquid vaporizes with minimal heat which prevents oxidation of samples during the drying process. High performance vacuum pumps ensure the low pressures which are required for faster solvent removal.
Heat is applied to the samples to accelerate the evaporation process. SpeedVac Vacuum Concentrators use built-in chamber heaters. Some models are equipped with radiant lamp on the lid to provide additional heat and prevent condensation of the solvents.
Virtual tour
Take a 3D tour and explore the Savant SpeedVac Integrated Vacuum Concentrator System
SpeedVac 3D Tour
Note: This tool is best viewed in Chrome, Firefox or Safari Browsers. Learn more ›
Smart Note: Savant SpeedVac System
When the quality of your results hinges on the quality of synthesized oligos
As oligos are being aggressively leveraged in research and clinical applications such as detecting, characterizing, and developing therapeutics and vaccines, the quality of synthesized oligos is critical. The Savant SpeedVac system enhances speed and reliability so you can have complete confidence in the quality of your oligos as well as the applications they serve.
Download
Featured information
Sample Evaporation with Thermo Scientific Savant SpeedVac Vacuum Concentrators
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Watch the video to learn about effective means of sample preparation.
      Questions to consider when selecting a SpeedVac system
      We have a lot of options available to anyone wanting to concentrate samples using the SpeedVac system. If you are confused as to which one is right for you, review the six questions below to learn more about which SpeedVac is right for your needs.
      Question #1: What is the material or solute that you are concentrating or drying?
      Is it biological or non-biological? We generally break solutes into two categories–biological (DNA, RNA, enzymes, proteins, vaccines, etc.) and non-biological (products of organic synthesis, drug metabolites, pesticides).
      The sample concentration and drying technique of choice for non-biological materials is vacuum evaporation. These samples are generally in high concentrations of organic solvents and/or strong acids and bases. Vacuum concentration offers rapid concentration of non-biological materials. The main question with biologicals is whether to freeze dry (lyophilize) or vacuum concentrate. Freeze drying is generally recommended when the material is to be stored for long periods of time, must be easily and quickly reconstituted, and needs to retain biological activity (i.e., proteins/enzymes). Most molecular biology applications involving DNA/RNA, in aqueous or aqueous + low concentration organics, use vacuum concentration for sample prep. Consider systems such as the Thermo Scientific™ Savant™ SpeedVac™ DNA130 or SPD1030.
      Freeze drying can be performed efficiently in a vacuum concentrator. However, freeze drying requires a deep vacuum (i.e., rotary vane oil pump) to keep the sample frozen during sublimation. Depending on the solute/ solvent and conditions, a properly configured SpeedVac system can be used for either freeze drying or vacuum concentration such as the SpeedVac SPD120P2.
      Question #2: What is the solvent that your material is prepared in?
      If it is an organic solvent, is it an aggressive solvent (methylene chloride, toluene), or a non-aggressive solvent (ethanol, methanol, acetonitrile) or a high-boiling-point solvent (DMSO, DMF)?
      This is one of the most important decision making factors in the proper choice of a SpeedVac system. Our goal is to give you the most cost-effective solution to your sample prep needs. Due to the need to withstand an increasingly harsh environment, the cost of a SpeedVac concentrator increases with the harshness of the solvent as follows:
      Aqueous < Non-aggressive organics < Aggressive organics < Strong acids/bases
      A SpeedVac system designed for use with strong acids/ bases would be suitable for use with other solvents such as non-aggressive organics, aggressive organics or aqueous. Consider systems such as the Thermo Scientific Savant SpeedVac SPD140DDA P1 or PSD300DDA P1. However, if you were only using aqueous or low concentration non-aggressive organics, you may not want to look at systems such as Thermo Scientific Savant SpeedVac SPD1030 or SPD2030.
      By considering your solvent(s) to be evaporated, you can select the SpeedVac that will meet your needs most cost effectively. (Be sure to consider your future needs; see also question 6.)
      Question #3: What type of vessels will be used? What is the sample volume and the number of samples to be processed?
      Thermo Scientific Savant offers SpeedVac systems and rotors capable of processing a wide variety of sample volumes and containers. Determine your format, working volume and number of tubes/plates or flasks, and this will help determine which size of system meets your needs; large capacity or small capacity.
      Question #4: Do you want a fully integrated system or a modular system?
      Thermo Scientific Savant pioneered integrated technology for vacuum concentration. Our fully integrated SpeedVac systems feature all components integrated into one compact system, optimized for performance, and factory tested to ensure your system is free of vacuum leaks. You take it out of a box, plug it in and you’re done. Integrated systems include the DNA130 and SPD1030.
      The only potential disadvantage of the integrated systems is flexibility to upgrade as your needs in the lab changes. Modular systems flexible design allows you to change components of the system should you need to add a secondary cold trap or different vacuum pump as the labs applications changes. A component system can be easily upgraded. Savant SpeedVac kits are available to make it easy to order a complete module system such as the SPD120P2, SPD210DXL, DNA140DDAP1 and SPD300DDAP1 include SpeedVac system, cold trap, vacuum pump and rotor.  
      Question #5: Do you want oil-free or oil vacuum pump technology?
      Any component SpeedVac system can be configured with either an oil-free diaphragm pump or rotary vane oil pump. We generally recommend oil-free technology for all applications except freeze-drying (which requires the deeper vacuum achieved with an oil pump) and evaporation of high-boiling-point solvents such as DMSO. The benefits of oil-free technology are numerous including ease-of maintenance and elimination of the cost of oil, oil filters, disposal of contaminated oil and pump rebuilds due to poor maintenance (i.e., “Who forgot to check the oil?”).
      The initial cost of an oil pump can be less than an oil-free pump…an important buying consideration. However, over the life of the pump, oil-free technology is the most cost effective, low maintenance choice.
      Question #6: What are your future application needs?
      Think about not only your current application needs but those needs that you may have in the future. For example, if you are currently using only aqueous solvents but foresee the need in the future to use aggressive solvents, purchase a system that will allow you to do both.
        Learn about special limited-time Lab Equipment offers.
        Go to the Lab Shop ›
      Support
      Unity Lab Services: Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.