1x1 image pixel for data collection
Visibly simple food pathogen and quality PCR testing - simple workflows designed to get results
Rapidly and accurately detect relevant foodborne pathogens and other important food safety and quality targets with the Thermo Scientific SureTect Food Safety PCR System. 
Empower your operations with simple, scalable workflows and rapid reporting to enable faster product release and interventions. Our food safety PCR assays offer detection of the major pathogens and other important targets in food, beverage, pet food and animal feed, primary production samples, and environmental samples.
Whether your laboratory is looking to introduce PCR technology for the first time, or you want to expand the PCR testing services you offer, we can help you find the right solution.  Benefit from:
Case study: Bringing the benefits of PCR and automation to high-throughput testing with the SureTect Automation Platform
Streamlined workflow for reliable results
rapid pathogen detection microorganisms validation  meat poultry salmonella research pathogens research use bacteria
Single enrichment step
8-24 hours
Single enrichment step of 8-24 hours (depending on target and matrix)
foodborne pathogens contaminated food poisoning pcr technology e coli food safety organism listeria contribute present
Sample preparation and lysis
20-40 minutes
Fully automated using the SureTect Automation Workflow or manual preparation with direct lysis in the Applied Biosystems SimpliAmp Thermal Cycler
foodborne illness occurs false positives vast majority healthy people listeria spp reliable results research use pathogens detection patients
Run, read, and report
~80 minutes
Perform PCR with the Applied Biosystems QuantStudio 5 Instrument using the RapidFinder Analysis Software. Report PCR-negative results immediately (results are automatically interpreted)
relevant foodborne pathogens  bacterial pathogens various applications symptoms contribute present detected  diagnosis
Confirm PCR positives
24 hours
Confirm PCR-positive results, after overnight incubation, using rapid chromogenic culture media methods
SureTect Features
Streamlined enrichment
Single versus dual enrichment protocols for faster time-to-result and streamlined workflow
develop laboratories pages microorganisms production poultry infections specificity detected outbreak range symptoms patients viruses research use samples cronobacter spp infectious disease food producers environmental samples
Validated assays
A range of assays validated for a broad range of targets and matrices according to certification requirements of AOAC and ISO 16140-2:2016 (by AFNOR or MicroVal)
contamination human risk pathogens listeria identification salmonella bacteria related species environmental samples food safety e coli
Flexible setup
Universal PCR conditions for detecting multiple targets in the same run
contamination human risk pathogens listeria identification salmonella bacteria related species environmental samples food safety e coli
Automatable workflows
Fully automated lysis and PCR setup with the Thermo Scientific SureTect Automation Platform
infectious disease pcr technology e coli environmental samples cronobacter spp samples  various applications symptoms
Intuitive instrumentation
‘Plug and play’ ready-to-use instruments out of the box for rapid pathogen dection
listeria identification research specificity detected present species pcr viral microorganisms meat develop solution analysis
Smart software
Fully automated amplification, detection, data collection, and data analysis using Thermo Scientific RapidFinder Analysis Software v3.0 or later
relevant foodborne pathogens food poisoning e coli sample preparation environmental samples reliable results detection salmonella
The SureTect PCR System – OMIC USA’s choice for reliable, robust foodborne pathogen detection 
OMIC case study
After reviewing several PCR solutions for foodborne pathogen testing, OMIC selected the Thermo Scientific SureTect PCR System for its laboratories' pathogen testing, favoring its robust, efficient workflow, compatibility with a wide range of matrices, and strong performance during validation.
Visible selective enrichment of Salmonella and Cronobacter species
PrecisBlue
In your busy Salmonella and Cronobacter testing workflows, there is no time for error. A common mistake, particularly in high throughout laboratories, is the accidental omission or duplication of adding a selective supplement during sample preparation.  For clearer selective enrichment of food samples, the new Thermo Scientific Oxoid PrecisBlue Supplement has been specifically designed to reduce workflow errors. The addition of an inert blue dye provides a clear visual reference, enabling easy identification of supplemented samples.
For Research Use Only. Not for use in diagnostic procedures.