Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031144.3 | NP_112406.1 | 4 - 5 | 881 | 91 | |
GenBank mRNA | AB028846.1 | - | 1 - 2 | 90 | 91 | |
BC063166.1 | - | 4 - 5 | 884 | 6920838 | 91 | |
EF156276.1 | - | 2 - 3 | 603 | 91 | ||
FQ226945.1 | - | 155 | 91 | |||
FQ227027.1 | - | 443 | 91 | |||
FQ227201.1 | - | 240 | 91 | |||
FQ229293.1 | - | 242 | 91 | |||
FQ231277.1 | - | 237 | 91 | |||
FQ231503.1 | - | 4 - 5 | 883 | 91 | ||
FQ231691.1 | - | 231 | 91 | |||
FQ232063.1 | - | 4 - 5 | 883 | 91 | ||
FQ232650.1 | - | 4 - 5 | 883 | 91 | ||
FQ232682.1 | - | 423 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Endogenous Controls Detect Genomic DNA Recommended Assay Rattus norvegicus Rat Androgen Panel Rat Chemokine Panel Rat Estrogen Panel Rat Tumor Metastasis Panel Rat Endogenous Controls 1700 Inventoried Rat_Endogenous_Controls Probe spans exons |