Vanquish Flex UHPLC Systems
For analyses that require more flexibility to accommodate all applications, Thermo Scientific Vanquish Flex UHPLC systems enable you to increase your productivity and drive innovation without compromising quality. Whether you need an LC or LC-MS solution, the flexibility of the Vanquish Flex systems with binary or quaternary solvent blending provides the most advanced performance through better separations, more results and easier interaction. Like all members of our Vanquish product line, Vanquish Flex UHPLC systems offer hardware precision, detector sensitivity, and simplicity of operation.
Build your ideal HPLC or UHPLC with our step-by-step configurator
Highlights
Separations with more flexibility
Thermo Scientific SmartFlow pumping technology includes automatic compensation for changes in eluent compressibility to ensure flow and gradient precision, independent of eluent composition and backpressure. The pump can operate up to 1000 bar and is available in binary, quaternary, and dual-gradient options.
Accurate sample handling
Thermo Scientific Vanquish Split Sampler FT offers outstanding speed and injection precision using Thermo Scientific SmartInject technology. It has a default vial capacity of 216 samples, which can be extended up to 8832 samples with the Thermo Scientific Vanquish Charger sampler extension.
Intelligent system tools
Two optional components are available to improve productivity, ensure compliance, and keep laboratories running smoothly. The Vanquish Solvent Monitor tracks solvent levels to prevent dry solvent channels and waste spills. Meanwhile, the touchscreen Vanquish User Interface provides visual monitoring of the system status.
Versatility in analyte detection
A variety of detection options ensure you can collect the most information from your separation. Our portfolio includes diode array detection, near-universal charged aerosol detection, fluorescence detection, mass spectrometric detection and more.
Biocompatible, easy and reliable tool-free connections
The fluidic pathway of the Vanquish Flex UHPLC system is fully biocompatible and fitted with Thermo Scientific Viper fingertight fittings, which support near-zero dead volume connections and enable sharper peaks and tool-free setup.
CDS software
The Vanquish Flex UHPLC system integrates with Thermo Scientific Chromeleon Chromatography Data System (CDS) with compliance tools, networking capabilities, automation, and data processing. We offer standard instrument integration (SII) packages to connect Thermo Scientific HPLC systems to other CDS and MS software programs.
Videos
System specifications
The table below provides an overview of the Vanquish Flex UHPLC system specifications for a standard setup. Not all specifications are listed. Your specifications may differ based on the components of your system. Find the specification sheets for the system components in the Component specifications, below.
Category Vanquish Flex UHPLC Systems
System pressure, maximum 1000 bar
Flow range, maximum 0–8 mL/min
Pump options (default gradient delay volume)
Binary (200 µL)
Quaternary (679 µL) 
Delay volume dependent on pump(s) selected
Biocompatibility Yes
pH range 2–12, chloride concentration up to 1 mol/L
System design Modular
Touchscreen user interface display compatible (optional) Yes
Solvent monitoring system compatible (optional) Yes
Customizable gradient delay volume to simplify method transfer Yes, with metering device
SmartInject technology for maximum injection precision and improved column lifetime Yes
Injection volume range
Standard: 0.01 - 25 µL
Optional: 0.01 - 100 µL
Sample cooling 4-40°C
Sample capacity (standard)
Based on standard 12 mm OD vials (≤1.5 mL)
4 racks of 54 vials (≤ 1.5 mL) each, other rack formats available
+ capacity of 12 × 22.5 mm OD vials (≤10 mL) in the carousel
Sampler extension available (optional)
Yes, with Vanquish Charger
Column compartment thermostatting Yes, 5-120°C 
Still air and forced air
Column compartment pre-heater Active and passive  
Column compartment post-column cooler Yes
Detector choices
Vanquish Charged Aerosol Detector
Vanquish Diode Array Detector
Vanquish Fluorescence Detector
Vanquish Multiple Wavelength Detector
Vanquish Variable Wavelength Detector
Refractive Index Detector
Mass Spectrometer
Component specifications
Standard Vanquish Flex UHPLC systems are made up of the following components. Download the associated specifications sheet for more detail on each component.
  Vanquish Flex Binary UHPLC Vanquish Flex Quaternary UHPLC
Pump
Download Specification Sheet
Vanquish Binary Pump F Vanquish Quaternary Pump F
Autosampler
Download Specification Sheet
Vanquish Split Sampler FT
Extended sample capacity (optional)
Download Specification Sheet
Vanquish Charger (optional)
Column compartment
Download Specification Sheet
Vanquish Column Compartment H
Virtually zero dead volume system connections
Download Specification Sheet
Viper UHPLC Fittings Kits
Detectors
Visit our HPLC and UHPLC Detectors page
Optional components: Fraction Collector Visit our Vanquish Fraction Collector FT page
Contact us to get a quote or discuss an HPLC/UHPLC solution
For Research Use Only. Not for use in diagnostic procedures.