Gas and Liquid Density Measurement
Minimize process interruptions
Reliably measure density or density related variables within the oil and gas, petrochemical, and power industries with Thermo Scientific Sarasota gas and liquid density meters. Our meters offer near real-time control signal availability and ensure plant efficiency and vitality.
Gas density measurement products
The composition of any gas is often characterized by its density or specific gravity. Thermo Scientific Sarasota gas density meters provide continuous, online monitoring of the density, or density related variables, to instantly detect any variation of process constituents or final product quality. The results are improved productivity, minimized product waste and reduced costs by providing real-time control as opposed to other sampling methods.
Use the Thermo Scientific Sarasota RTR900 Instrument Retractor with the Thermo Scientific Sarasota ID900 gas density meter in high-pressure measurement applications. This screw jack allows insertion and removal of the ID900 under operational line conditions, allowing measurement at true process conditions.
Measure the density, pressure and temperature of dry and wet gases with the Thermo Scientific Sarasota SG901 (formerly SG900) Specific Gravity Analyzer, a self-contained bypass sample conditioner. The compact model features fast response and continuous, high accuracy measurements to 0.2% of reading, making it an ideal choice for a variety of applications.
Achieve optimum data processing with Thermo Scientific CM515 and HME900 Density Converter Electronics. Thermo Scientific density meters or specific gravity analyzers are available with either a HART compatible, field-mounted density converter or a control room-based unit. Both instruments provide high-integrity calculations of density and density-related variables to ensure effective custody transfer and process optimization.
Liquid density measurement
Thermo Scientific Sarasota liquid density meters utilize the proven vibrating element design which is widely accepted as the most accurate method of continuous, online density measurement. Compact and lightweight, the density meters tolerate significant plant vibration and can be installed directly into existing pipe work without the need for upstream flow conditioning or instrument supports.
Achieve optimum data processing with Thermo Scientific CM515 and HME900 Density Converter Electronics. Thermo Scientific density meters or specific gravity analyzers are available with either a HART compatible, field-mounted density converter or a control room-based unit. Both instruments provide high-integrity calculations of density and density-related variables to ensure effective custody transfer and process optimization.
environmental monitoring product support and spare parts
Product support and spares
Thermo Fisher Scientific Service Solutions are a vital component of your product’s lifecycle. These comprehensive solutions are exclusively created to drive optimal performance of your time-critical instruments.
When you partner with Thermo Fisher Scientific, you gain access to engineers trained within the same factory your instruments were manufactured. We only ship spare parts that were designed specifically for your instrument. Our expert service team is truly an integrated part of your product’s life cycle.
CMD SchemaApp code