Sulfur Analyzers
Reliable total and trace sulfur analyzers
Online sulfur analyzers replace expensive and time-consuming laboratory sampling. Get fast responses and wide measurement ranges of trace sulfur and total sulfur in flare gas, liquid and vapor for reliable emissions monitoring.
SOLA iQ Applications
Clean Fuels
Clean Fuels
The superior stability and precision of the SOLA iQ enables refiners to make timely process adjustments to enhance the economic efficiency of desulfurization and fuel blending operations.
  • SOLA iQ Liquid
  • SOLA iQ Liquid Trace
Flare Gas & Condensable Vapors
Flare Gas & Condensable Vapors
The highly accurate SOLA iQ Flare analyzer features a dynamic measuring range from 10 ppm to 100% S by volume with fast high-to-low response time, enabling reliable flare stack sulfur emission reporting. The SOLA iQ Condensible Vapors measures 5ppm to 1% sulfur and trace sulfur.
  • SOLA iQ Condensible Vapors (CV)
  • SOLA iQ Condensible Vapors (CV) Trace
  • SOLA iQ Flare
Multi-Calibration/Multi-Stream
Multi-Calibration/Multi-Stream
The SOLA iQ analyzer enables multiple streams of different sulfur concentrations (i.e., batch processing, inlet/outlet of reactors, etc.) to be measured by a single analyzer.
  • SOLA iQ Liquid
  • SOLA iQ Vapor
  • SOLA iQ Condensible Vapors (CV)
  • SOLA iQ Flare
  • SOLA iQ Trace
Clean Fuels
Clean Fuels
The superior stability and precision of the SOLA iQ enables refiners to make timely process adjustments to enhance the economic efficiency of desulfurization and fuel blending operations.
  • SOLA iQ Liquid
  • SOLA iQ Liquid Trace
Flare Gas & Condensable Vapors
Flare Gas & Condensable Vapors
The highly accurate SOLA iQ Flare analyzer features a dynamic measuring range from 10 ppm to 100% S by volume with fast high-to-low response time, enabling reliable flare stack sulfur emission reporting. The SOLA iQ Condensible Vapors measures 5ppm to 1% sulfur and trace sulfur.
  • SOLA iQ Condensible Vapors (CV)
  • SOLA iQ Condensible Vapors (CV) Trace
  • SOLA iQ Flare
Multi-Calibration/Multi-Stream
Multi-Calibration/Multi-Stream
The SOLA iQ analyzer enables multiple streams of different sulfur concentrations (i.e., batch processing, inlet/outlet of reactors, etc.) to be measured by a single analyzer.
  • SOLA iQ Liquid
  • SOLA iQ Vapor
  • SOLA iQ Condensible Vapors (CV)
  • SOLA iQ Flare
  • SOLA iQ Trace
Determine total sulfur and trace sulfur concentration in liquid or gas phase samples to ensure process optimization, maximize profitability, and maintain regulatory compliance. Utilizing unique Pulsed Ultraviolet Fluorescence (PUVF) technology and the proven Pyrolysis system, the SOLA iQ analyzer builds on the success of the SOLA II range of total sulfur analyzers and measures from 2ppm to 100% total sulfur content, with detection as low as 25ppb.
View SOLA iQ FAQ
Perform total sulfur analysis on liquid hydrocarbon fuel process streams with the Thermo Scientific SOLA II Sulfur Online Analyzer. This proven field sulfur analyzer addresses the need for a low-maintenance total sulfur analysis instrument that can reliably measure low ppm levels to help customers meet the tightening, Tier 3, regulatory requirements that limit sulfur levels in fuels.
HDI-AutoConfig-Software-270x195
Thermo Scientific AutoCONFIG software is the robust interface of the SOLA iQ.  Designed to empower the user, the interface is simple to configure eliminating the need for in depth programming.
environmental monitoring product support and spare parts
Product support and spares
Thermo Fisher Scientific Service Solutions are a vital component of your product’s lifecycle. These comprehensive solutions are exclusively created to drive optimal performance of your time-critical instruments.
When you partner with Thermo Fisher Scientific, you gain access to engineers trained within the same factory your instruments were manufactured. We only ship spare parts that were designed specifically for your instrument. Our expert service team is truly an integrated part of your product’s life cycle.
CMD SchemaApp code