Our gas isotope ratio mass spectrometers are the instruments of choice for measuring stable isotope signatures with extreme precision and sensitivity, enabling the user to gain unique insights into the history and origin of compounds found in a variety of samples.
Whether you need flexibility to perform IRMS applications in one single analyzer, or you need robustness, stability and precision for high performance IRMS, or whether you need sensitivity and high resolution for unraveling the isotopic anatomy of molecules, our gas IRMS product portfolio offers it all. In combination with our unique sample preparation devices and inlet systems, our gas isotope ratio mass spectrometers offer you a comprehensive instrumental setup for your applications.
Perform all IRMS applications in a single analyzer
DELTA Q Isotope Ratio Mass Spectrometer
The Thermo Scientific DELTA Q Isotope Ratio Mass Spectrometer combines outstanding sensitivity with excellent linearity and stability to address diverse applications.
Learn more about DELTA Q IRMS ›
Get the highest sensitivity and performance
253 Plus 10 kV Isotope Ratio Mass Spectrometer
The Thermo Scientific 253 Plus 10 kV Isotope Ratio Mass Spectrometer provides a flexible and open platform for the connection of inlet systems and preparation devices for smallest amounts of sample in IRMS.
Learn more about 253 Plus 10 kV IRMS ›
Explore the world in a whole new way
Ultra High Resolution Isotope Ratio Mass Spectrometer (HR-IRMS)
With its guaranteed mass resolving power of 30,000 (5%, 95% edge definition), the Thermo Scientific Ultra High Resolution IRMS accurately determines conditions by which molecules were formed, transported, stored, and degraded.
Learn more about Ultra High Resolution IRMS ›
High-resolution IRMS
Mass resolving power greater than 18,000
The Ultra HR-IRMS combines a 10 kV electron impact ionization source, a high-mass-resolution double-focusing magnetic-sector mass analyzer, and a variable collector array for a resolving power of 18,000. A high, medium, or low resolution setting for each method can automatically be selected for by using a switchable entrance slit.
Ultimate abundance sensitivity
The variable collector array covers a signal dynamic range greater than 10 orders of magnitude, facilitating the collection of more abundant ion beams in Faraday cups as well as single ion counting of very rare ion species. Weak signals can be analyzed with the axial secondary electron multiplier using the retarding potential quadrupole (RPQ) retardation lens, enabling experiments of high sensitivity.
Fully integrated workflows
The Thermo Scientific Qtegra Intelligent Scientific Data Solution (ISDS) revolutionizes the steps involved in isotope ratio analysis with fully integrated workflows, making high-resolution IRMS a routine tool. Qtegra ISDS Software combines all experiments performed on a specific sample into one "lab book."
Modular concept
The Ultra HR-IRMS includes two dual viscous flow inlet systems for handling up to three reference gases together with the sample. This modular dual inlet design allows each dual inlet to be exchanged, avoiding cross contamination of the sample types being analyzed.
High resolution gas IRMS components
Electrostatic detector
The electrostatic sector analyzer (ESA) with a radius of 220mm refocuses the energy dispersion of the ions prior to the magnetic sector.
Entrance slit
The computer-controlled entrance slits can be switched between 5 μm and 250 μm which allows the selection of low, medium, and high mass resolution.
Magnetic sector
The temperature-controlled, laminated electromagnet has a radius of 230 mm. Overall system magnification is a factor of 2, yielding a dispersion of 810 mm. Mass range is up to mass 280 at full acceleration voltage, extendable to 560 at reduced accelerating potential and mass resolving power.
Variable multicollector
On either side of the fixed axial collector there are 4 moveable detector platforms. Each variable detector platform is equipped with a Faraday detector and in addition can carry a compact discrete dynode (CDD) ion counting detector.
10 kV ion source
The gas-tight electron impact ion source with a wide range of adjustable electron energy and variable pumping conductance provides ultra high sensitivity. The ion source preserves clumping and isotopic distribution in the ions because of its low “scrambling” characteristics.
RPQ SEM
The axial detector channel is a dual mode detector with a Faraday cup and a high sensitivity ion counting detector (SEM). In front of the SEM is a retarding potential quadrupole (RPQ) to reduce the abundance sensitivity for measurement of very small ion beams.
Chemical and physical processes leave traces in the isotopic fingerprint of samples. Nitrogen, carbon, sulfur, oxygen, and hydrogen preserve the information related to those processes in their isotope ratios. Our comprehensive portfolio of gas IRMS peripherals helps you access unique sample information and gain conclusive insights in food and beverage testing, forensics, archaeology, ecology, and geosciences.
EA-IRMS: Reach higher chromatography peaks 
With the Thermo Scientific EA IsoLink IRMS System, laboratories have an automated, easy-to-use solution for elemental and isotopic analysis. With dedicated helium-saving technology and integration of temperature ramped gas chromatography, analyses are fast and cost-efficient for origin and authenticity determination of samples in a variety of fields: ecology, biology, archaeology, geosciences, food integrity, and forensics.
 View EA IsoLink schematic
Learn more
Powerful GC-IRMS separations
The Thermo Scientific GC IsoLink II IRMS System creates a seamless solution that combines the separation capabilities of GC with the superior detection power of IRMS. In addition to providing scientists with a wealth of information on compound origin, metabolic, synthetic and diagenetic pathways, the GC IsoLink II IRMS System can be connected with GC/GC-MS techniques to reveal unknown compounds.
 View GC IsoLink schematic
Learn more
LC-IRMS for specific compounds from bulk liquid samples
The Thermo Scientific LC IsoLink II IRMS System is the next generation LC-IRMS platform allowing sensitive, precise determination of carbon isotope ratios of polar compounds and bulk samples. All-in-one, integrated stack design features the innovative Thermo Scientific Vanquish Core HPLC for a variety of applications such as food integrity, metabolism and physiology studies, drug origin verification, forensics, archaeology, doping control and biogeochemistry research.
 View LC IsoLink II schematic
Learn more
GasBench Plus System: Flexibility, sensitivity, dynamic range
The Thermo Scientific GasBench Plus System fits a diverse range of applications, from natural isotope variation to beverage authenticity and metabolism testing. Automated range recognition and autodilution combine with a dual viscous flow inlet system, resulting in precision, sensitivity, and a large dynamic range, as well as fast delivery of pure analyte from complex gas mixtures.
 View GasBench schematic
Learn more
Kiel IV Carbonate Device
When coupled to an isotope ratio mass spectrometer, the Thermo Scientific Kiel IV Carbonate Device enables analysis of carbonate samples for paleoclimatic reconstructions involving biogenic carbonates such as foraminifera, bivalves, brachiopods, otoliths, and corals. Other applicable fields of study include analysis of growth zones of individual organisms or microfossils from drill cores.
Learn more
ConFlo IV Universal Continuous Flow Interface
The Thermo Scientific ConFlo IV Universal Continuous Flow Interface enables attachment of sample preparation devices to the IRMS that dramatically expand the range of experiments that can be conducted by stable IRMS.
Learn more
EA-IRMS: Reach higher chromatography peaks 
With the Thermo Scientific EA IsoLink IRMS System, laboratories have an automated, easy-to-use solution for elemental and isotopic analysis. With dedicated helium-saving technology and integration of temperature ramped gas chromatography, analyses are fast and cost-efficient for origin and authenticity determination of samples in a variety of fields: ecology, biology, archaeology, geosciences, food integrity, and forensics.
 View EA IsoLink schematic
Learn more
Powerful GC-IRMS separations
The Thermo Scientific GC IsoLink II IRMS System creates a seamless solution that combines the separation capabilities of GC with the superior detection power of IRMS. In addition to providing scientists with a wealth of information on compound origin, metabolic, synthetic and diagenetic pathways, the GC IsoLink II IRMS System can be connected with GC/GC-MS techniques to reveal unknown compounds.
 View GC IsoLink schematic
Learn more
LC-IRMS for specific compounds from bulk liquid samples
The Thermo Scientific LC IsoLink II IRMS System is the next generation LC-IRMS platform allowing sensitive, precise determination of carbon isotope ratios of polar compounds and bulk samples. All-in-one, integrated stack design features the innovative Thermo Scientific Vanquish Core HPLC for a variety of applications such as food integrity, metabolism and physiology studies, drug origin verification, forensics, archaeology, doping control and biogeochemistry research.
 View LC IsoLink II schematic
Learn more
GasBench Plus System: Flexibility, sensitivity, dynamic range
The Thermo Scientific GasBench Plus System fits a diverse range of applications, from natural isotope variation to beverage authenticity and metabolism testing. Automated range recognition and autodilution combine with a dual viscous flow inlet system, resulting in precision, sensitivity, and a large dynamic range, as well as fast delivery of pure analyte from complex gas mixtures.
 View GasBench schematic
Learn more
Kiel IV Carbonate Device
When coupled to an isotope ratio mass spectrometer, the Thermo Scientific Kiel IV Carbonate Device enables analysis of carbonate samples for paleoclimatic reconstructions involving biogenic carbonates such as foraminifera, bivalves, brachiopods, otoliths, and corals. Other applicable fields of study include analysis of growth zones of individual organisms or microfossils from drill cores.
Learn more
ConFlo IV Universal Continuous Flow Interface
The Thermo Scientific ConFlo IV Universal Continuous Flow Interface enables attachment of sample preparation devices to the IRMS that dramatically expand the range of experiments that can be conducted by stable IRMS.
Learn more
Qtegra ISDS
Qtegra ISDS Software for gas IRMS
Minimize training, automate workflows, simplify your experience, and improve efficiency with the innovative Qtegra Intelligent Scientific Data Solution (ISDS) platform software. Designed for simplified workflow compliance, scalability, compliance and data management, Qtegra ISDS Software provides essential tools for consistent, accurate analysis.
Learn more about Qtegra ISDS Software ›
Qtegra ISDS Software for gas IRMS webinar: revolutionize your gas IRMS analyses ›
Isotope fingerprinting
Scientific research
Archaeology
Forensics
Biology & Biogeochemistry
Oil & Gas
Climate change research
Partnering in sustainability - ACT label
Reduce your lab’s environmental impact with the DELTA Q IRMS, recipient of the ACT label from My Green Lab. The ACT label, the standard for determining the environmental impact of laboratory products, helps labs make informed, sustainable purchasing decisions.
Learn more about the ACT label ›
Featured videos
Tracing origin of pollution with the Isotope Hunter
To protecting our environment from contamination you need to identify the source. By tracing Carbon, Oxygen, Hydrogen, Nitrogen, and Sulfur Isotope Fingerprint, the Isotope Hunter reveals clues that are invaluable to Environmental investigations.
Video Player is loading.
Current Time 0:00
Duration 4:27
Loaded: 0%
Stream Type LIVE
Remaining Time 4:27
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
      1. Now Playing
        Up NextTracing origin of pollution with the Isotope Hunter
        To protecting our environment from contamination you need to identify the source. By tracing Carbon, Oxygen, Hydrogen, Nitrogen, and Sulfur Isotope Fingerprint, the Isotope Hunter reveals clues that are invaluable to Environmental investigations.
      2. Now Playing
        Up NextTracing pollution Sources in Air with Isotope Fingerprints
        Isotope Fingerprints help trace pollution sources, enabling the monitoring of water and air quality and supporting studies on environmental changes.
      3. Now Playing
        Up NextDetecting Wine Adulteration with Isotope Fingerprints
        Isotope Fingerprints are used to detect addition of water or juices to wine with the aim of ensuring consumer trust and fairer market competition
      4. Now Playing
        Up NextTracing human provenance using Isotope Fingerprints
        Isotope fingerprints provide important additional evidence that has been able to trace human movements and bring clarity to homicide cases.
      5. Now Playing
        Up NextIsotope Fingerprints in Authenticity and Food Fraud
        Isotope fingerprints are powerful tools to determine authenticity, country of origin and potential frauds for food and beverage samples.
      6. Now Playing
        Up NextDoping Control with the Isotope Hunter
        With isotope fingerprints and IRMS you’ll discern their true identities, quantities, origins, and, most importantly, their legality under World Anti-Doping Agency regulations.
      7. Now Playing
        Up NextMeet the Isotope Hunters at Eurofins, Nantes
        The Isotope Hunters at Eurofins, Nantes investigate, also with isotopes, on food authenticity, to ensure consumer trust and fairer market competition
      8. Now Playing
        Up NextWelcome to the Isotope Hunter Lab
        Tracking origins and tracing identity: the Isotope Hunter follows the clues that history leaves behind in food or fiber, liquid or stone. Capture your samples’ stories with Thermo Scientific’s portfolio of IRMS.
      9. Now Playing
        Up NextMeet the Isotope Hunters at Imprint Analytics
        By using isotope fingerprints the isotope Hunters at Imprint Analytics fight against Food fraudsters to protect consumers.
      10. Now Playing
        Up NextInvestigating criminal forensic cases with the Isotope Hunter
        With crime comes evidence. When an investigation hits a wall it’s time to call on the Isotope Hunter.
      11. Now Playing
        Up NextInvestigating authenticity and tracing origin with isotope fingerprints
        Food and beverage samples have a fingerprint, a unique chemical signature that allows the product to be identified with IRMS.
      12. Now Playing
        Up NextVerifying Food Origins Using Stable Isotopes
        Dr Simon Kelly explains how isotope analysis is used for the determination of adulteration cases in food and beverages, with examples on honey.
      13. Now Playing
        Up NextUse of stable isotope analysis in commercial food authenticity testing
        New isotopic tools for testing geographic origin of food and naturalness of food ingredients meet food industry needs for speed, versatility and quality.
      14. Now Playing
        Up NextIsotope Fingerprints in Authenticity and Food Fraud
        Isotope fingerprints are powerful tools to determine authenticity, country of origin and potential frauds for food and beverage samples.
      15. Now Playing
        Up NextIsotope fingerprints: Origin of tequila with GC coupled with Isotope Ratio Mass Spectrometry
        Isotope fingerprints analyzed by GC-IRMS enable to differentiate tequila purity and can inform on adulteration and product mislabeling.
      16. Now Playing
        Up NextIsotope Fingerprints: Authentication of Honey by LC Coupled with Isotope Ratio MS
        Isotope fingerprints analyzed by LC-IRMS enable to identify if the sugar in the honey is natural or was artificially added.
      17. Now Playing
        Up NextAuthenticity Control of Beverages and Water in food with GasBench II System using Isotope Fingerprints
        Oxygen isotope fingerprint analyzed with the Thermo Scientific GasBench II System allows the identification of water addition in commercial wine.
    * Required field
    *
    *
    *
    *
    *
    *
    *
     Please confirm you would like to receive marketing and promotional email messages about Thermo Fisher Scientific products and services.
    This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
    CMD Form Code v3.0
    Style Sheet for Global Design System
    Style Sheet for Komodo Tabs
    CMD Wide-format style fixes