VISIONlite™ Spectrophotometer Software
VISIONlite™ Spectrophotometer Software
Thermo Scientific™
VISIONlite™ Spectrophotometer Software
Thermo Scientific™ VISIONlite™ 5 Software enhances the capabilities of our stand-alone Visible and UV-Visible spectrophotometers. The software enables you use your spectrophotometer to its fullest potential, displaying and reporting experimental and analytical results with ease.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
869-175600VISIONlite ColorCalc Software
840-312300VISIONlite 5 Software + Cable
869-191700Wine Analysis Software
869-191600VISIONlite EnzLab Software
Catalog number 869-175600
Price (USD)
-
Request A Quote
Type:
VISIONlite ColorCalc Software
Thermo Scientific™ VISIONlite™ 5 Software enhances the capabilities of our stand-alone Visible and UV-Visible spectrophotometers. The software enables you use your spectrophotometer to its fullest potential, displaying and reporting experimental and analytical results with ease.
Thermo Scientific™ VISIONlite™ ColorCalc software is a powerful stand-alone software package that adds the ability to perform numerous color calculations for a variety of applications. The software records the required transmission or reflectance spectrum and automatically performs calculations using the recorded data.
Thermo Scientific™ VISIONlite™ EnzLab software offers predefined methods and operation procedures for all standard enzymatic test kits including Citric Acid, Lactic Acid, D-Glucose, D-Fructose, Ethanol, Lactose and many more. EnzLab automates and facilitates enzymatic determinations, ensures correct calculations, and provides appropriate analytical reports.
Thermo Scientific™ VISIONlite™ Wine Analysis software allows users to perform an extensive list of predefined photometric, colorimetric and enzymatic analytical procedures common in wine and juice analysis. This PC software features a simple user interface, reliable data recording, and error-free evaluation of results within comprehensive reports.
VISIONlite Software enhancements include:
Scan:
  • Peak pick to identify and label wavelength peaks or valleys
  • Import, save or export data in native or CSV format for easy archiving and reporting
  • Auto scaling of data plus simple zoom controls for detailed examination of spectra 
Rate:  For SPECTRONIC 200 and GENESYS 30, measure multi-cell kinetics in parallel or serial mode
  • Automatic enzyme activity calculation
  • Automatic data storage plus import or export your data in CSV format
  • Measure up to 100 samples in a single data set 
Fixed:
  • Measure at one or multiple wavelengths
  • Apply factors and equations to your data to generate results directly
  • Addition, subtraction, ratio and baseline correction options
  • Run standard bio-tests using the preprogrammed protein and DNA methods
Quant: More standards and non-linear curve fitting options than can be accomplished under local control on Thermo Scientific SPECTRONIC 200 and GENESYS 30 Spectrophotometers
Specifications
For Use With (Equipment)Thermo Scientific GENESYS 30, 40, 50, 140, 150, 180, BioMate 160, Evolution 201, 220, 260 Bio, 350 Spectrophotometers, SPECTRONIC 200 and GENESYS 30
IncludesUSB with software installer and user manual (in English and German). Display languages: English, German, Spanish, Portuguese, French, Italian, Russian and Korean
Required AccessoriesInterface cable 840-312200 for Thermo Scientific GENESYS 40-180 and BioMate 160 Spectrophotometers. 085-820200 - Standard USB cable for Thermo Scientific GENESYS 30 Spectrophotometer.
TypeVISIONlite ColorCalc Software
Unit SizeEach