Search
Built to perform, the advanced Thermo Scientific TSQ Certis Triple Quadrupole Mass Spectrometer (MS) helps deliver streamlined, connected, and ready-to-run triple quadrupole workflows that keep your lab productive, profitable, and worry-free—no matter who’s at the bench.
With latest hardware for higher sensitivity and software for a compliant-ready solution, TSQ Certis Mass Spectrometer delivers greater uptime and improved performance for routine biopharmaceutical, clinical research, toxicology, environmental, and food safety testing laboratories.
Maximize throughput and efficiency in high-performance laboratories
Advanced ion optics lead to increased uptime, delivering high profitability for fee-for-service customers
Newly engineered OptaMax Plus Ion Source maintains stability and precision of measurement across a vast range of methods and sample types
Go from sample to results with speed, confidence and compliance
Decrease both energy usage and instrument maintenance with an environmentally-friendly dry pump option
Obtain world-class service solutions for all your service and support needs
The TSQ Certis Mass Spectrometer delivers accuracy you can defend, traceability you can audit, and reproducibility you can trust—across biopharmaceutical, clinical research, toxicology environmental analysis, and food safety testing high-throughput routine labs who must make critical, compliance-driven health decisions.
Analyzing human whole blood by LC-MS presents persistent challenges: matrix contaminants can accumulate within the ion source and API stack, compromising signal integrity and reproducibility. Restoring data quality demands time-consuming maintenance. The TSQ Certis Mass Spectrometer proves its durability by maintaining stable, precise performance through tens of thousands of injections—minimizing downtime and maximizing throughput.
Demand for highly sensitive, specific, and stable methods for the bioanalysis of GLP-1 peptides and oligonucleotides is increasing. The development of LC-MS assays for GLP-1 peptides and oligonucleotide therapeutics has advanced rapidly as biopharmaceutical programs increasingly demand highly selective, quantitative methods for pharmacokinetic (PK) and biomarker studies. Both molecule classes present complex analytical challenges—GLP-1 analogs due to their size, instability, and endogenous background; oligonucleotides due to their polarity, charge, and susceptibility to degradation.
By combining the Thermo Scientific Solaμ SAX SPE Extraction Plate with the TSQ Certis Triple Quadrupole Mass Spectrometer, laboratories can achieve exceptional sensitivity and specificity for GLP-1 peptides—reaching assay detection limits with greater confidence in target biomarker quantitation.
Liquid-liquid extraction of human plasma was used for quantitative bioanalysis of antisense oligonucleotides such as Fomivirsen and Nusinersen. The TSQ Certis Mass Spectrometer enables accurate measurement of oligonucleotides in challenging matrices such as plasma down to an LOQ of 0.1ng/mL.
Liquid chromatography coupled with triple quadrupole (LC–QqQ) mass spectrometry has become the industry standard for the quantitative determination of steroids in biological matrices. Compared to immunoassays, LC–QqQ MS offers superior analytical specificity, lower detection limits, and improved reproducibility, making it ideal for regulated pharmacokinetic, endocrinological, and clinical research applications. The TSQ Certis Mass Spectrometer provides the speed, specificity, and ultra-high sensitivity necessary for reliable steroid quantitation.
Our general purpose product lines, including LC-MS products, are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our general purpose product lines have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in health and safety risks. They are designated for Research Use Only Applications.
For Research Use Only - Not For Use In Diagnostic Procedures.