c7c8 - Key/Specs/Resources/Appl/Tech/Doc/Contact
Volumescope 2 Scanning Electron Microscope for serial block face imaging
The Thermo Scientific Volumescope 2 Scanning Electron Microscope (SEM) is our state-of-the-art serial block-face imaging system. Keep control of experiments with easy to use technology and protect a wide range of possible samples with tried and tested solutions for every acquisition step.
Block face imaging
Unraveling the complex 3D architecture of cells and tissues in their natural context is crucial for structure-function correlation in biological systems and soft materials. Serial block-face SEM (SBF-SEM) combines in situ sectioning and imaging of plastic-embedded tissue blocks within the SEM vacuum chamber for automated, 3D reconstruction of large tissue volumes. Axial resolution was once limited by the minimum section thickness, but truly isotropic resolution is now possible with the addition of multi-energy deconvolution.
3D SEM
Following in situ sectioning of the block-face using a diamond knife, the freshly exposed tissue is imaged several times using increasing accelerating voltages. These images are subsequently used in a deconvolution algorithm to derive several optical subsurface layers, forming a 3D subset. By repeating this cycle, the Volumescope 2 SEM offers isotropic datasets with 10 nm Z-resolution.
Mouse brain sample imaged on the Volumescope 2 SEM.
Mouse brain sample imaged on the Volumescope 2 SEM. Fine features such as vesicles and synaptic clefts can be observed. Image courtesy of Dr. Gabor Nyiri, Institute of Experimental Medicine, Budapest, Hungary.
Volume reconstruction of a mouse heart muscle.
Volume reconstruction of a mouse heart muscle. Data acquired by a combination of physical and virtual slicing in high vacuum mode. Volume = 16 x 15 x 11 μm3 with a nominal resolution of 5 x 5 x 10 nm. Specimen courtesy of Dr. Madesh Muniswamy, Temple University, USA.
Polymer microstructures
High-resolution imaging in 3D is of great importance when trying to understand the microstructure and properties of polymer materials. The Volumescope 2 SEM combines in situ sectioning and imaging of polymers within the SEM vacuum chamber, in a fully automated fashion, for reconstruction of large volumes with truly isotropic 3D resolution. Visualizing a large volume at such resolutions is critical for revealing how regions with different microstructures affect the overall properties of the material. For example, the Volumescope 2 SEM can recreate a large representative volume of a filtration membrane, providing accurate transport properties that lead to enhanced performance predictions for new filter designs.
Polymer blend imaged with Volumescope SEM.
Serial block-face 3D imaging of a polymer blend sample. Here, the 3D volume reconstruction of the complete dataset (122 μm x 122 μm x 10 μm) is shown, with a segmented region inset revealing the particle distribution in the matrix. Sample courtesy of Dr. Armin Zankel (FELMI-ZFE Graz, Austria).
Key Features
Easy to use technology
Keep control of experiments with easy to use technology. Reuse jobs and system settings and select multiple regions of interest during job acquisition.
Visualize and navigate during acquisition
Visualize and navigate during acquisition with Thermo Scientific Amira Live Tracker Software to optimize/control your outcome/Automate large 3D volume acquisitions as well as reconstructions.
Protect valuable samples
Protect valuable samples with tested solutions at every acquisition step: debris trap and swipe features ensure sample quality; low vacuum detector enables imaging of highly charged samples.
Easy and fast mounting microtome exchange
Easy and fast mounting microtome exchange for normal SEM operation or automated array tomography with the addition of Thermo Scientific Maps Software.
Specifications
Style Sheet for Products Table Specifications
Electron optics
  • High-resolution field emission SEM column
    • High-stability Schottky field emission gun
    • Compound final lens: a combined electrostatic, field-free magnetic and immersion magnetic objective lens
  • Dual objective lens combining electromagnetic and electrostatic lenses
Source lifetime
24 months
Automation
  • Auto bakeout
  • Auto start
  • No mechanical alignments
  • Automated heated apertures
  • User guidance and column presets
Stage
Double stage scanning deflection
Electron beam
  • Continuous beam current control and optimized aperture
  • Beam current range: 1 pA to 400 nA
    • In serial block-face imaging SEM: 50 pA – 3.2 nA
  • Landing energy range: 20 eV – 30 keV
  • Accelerating voltage range: 200 V – 30 kV
    • In serial block-face imaging SEM: 500 V – 6 kV
  • Resolution at optimum working distance and routine SEM high-vacuum imaging
    • 0.8 nm at 30 keV STEM
    • 0.7 nm at 15 keV
    • 1.0 nm at 1 keV
Chamber
  • Inside width: 340 mm
  • Analytical working distance: 10 mm
  • Ports: 12
Detectors
  • Serial block-face imaging detectors
    • T1 segmented lower in-lens detector
    • VS-DBS: LoVac lens-mounted BSED
  • T2 in-lens detector
  • Everhart-Thornley SE detector (ETD)
  • IR-CCD
  • Low-vacuum SE detector
  • STEM 3+ – Retractable segmented detector
  • Additional detectors available
Vacuum system
  • Complete oil-free vacuum system
  • 1 × 220 l/s TMP
  • 1 × PVP-scroll
  • 2 × IGP
  • Chamber vacuum (high-vacuum) <6.3 × 10-6 mbar (after 72 hours pumping)
  • Low-vacuum mode up to 50 Pa for charge compensation of non-conductive samples
  • Evacuation time: ≤3.5 minutes
Sample holders
  • Standard multi-purpose holder; uniquely mounts directly onto the stage; hosts up to 18 standard stubs (Ø12 mm), three pre-tilted stubs, two vertical and two pre-tilted row-bar holders
  • Each optional row-bar accommodates six S/TEM grids
  • Wafer and custom holders
See datasheet for full list of specifications including software, accessories and a summary of installation requirements.
Resources
Debris Management Cut
Klaus talks about the Debris Management.
Video Player is loading.
Current Time 0:00
Duration 0:19
Loaded: 0.00%
Stream Type LIVE
Remaining Time 0:19
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
      1. Now Playing
        Up NextDebris Management Cut
        Klaus talks about the Debris Management.
      2. Now Playing
        Up NextPerception of new UI workflow
        Klaus talks about the new user interface.
      3. Now Playing
        Up NextSBFI future
        Hear Klaus’ opinion about the future of SBFI.
      4. Now Playing
        Up NextSBFI - VS
        Key features
    Video Player is loading.
    Current Time 0:00
    Duration 0:41
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 0:41
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • default, selected
      3D volume visualization of the fracture zone of a polymer blend (iPP/EPR particles) after tensile test.
      Video Player is loading.
      Current Time 0:00
      Duration 2:40
      Loaded: 6.25%
      Stream Type LIVE
      Remaining Time 2:40
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • default, selected
        Serial block-face 3D volume imaging of a filter membrane using the Volumescope 2 SEM.
        Debris Management Cut
        Klaus talks about the Debris Management.
        Video Player is loading.
        Current Time 0:00
        Duration 0:19
        Loaded: 0.00%
        Stream Type LIVE
        Remaining Time 0:19
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
            1. Now Playing
              Up NextDebris Management Cut
              Klaus talks about the Debris Management.
            2. Now Playing
              Up NextPerception of new UI workflow
              Klaus talks about the new user interface.
            3. Now Playing
              Up NextSBFI future
              Hear Klaus’ opinion about the future of SBFI.
            4. Now Playing
              Up NextSBFI - VS
              Key features
          Video Player is loading.
          Current Time 0:00
          Duration 0:41
          Loaded: 0.00%
          Stream Type LIVE
          Remaining Time 0:41
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
            • default, selected
            3D volume visualization of the fracture zone of a polymer blend (iPP/EPR particles) after tensile test.
            Video Player is loading.
            Current Time 0:00
            Duration 2:40
            Loaded: 0.00%
            Stream Type LIVE
            Remaining Time 2:40
            1x
              • Chapters
              • descriptions off, selected
              • captions off, selected
              • default, selected
              Serial block-face 3D volume imaging of a filter membrane using the Volumescope 2 SEM.
              Applications
              Pathology research
              Pathology Research
              Transmission electron microscopy (TEM) is used when the nature of the disease cannot be established via alternative methods. With nano-biological imaging, TEM provides accurate and reliable insight for certain pathologies.
              Plant biology research
              Plant Biology Research
              Fundamental plant biology research is enabled by cryo electron microscopy, which provides information on proteins (with single particle analysis), to their cellular context (with tomography), all the way up to the overall structure of the plant (large volume analysis).
              Fundamental Materials Research_R&amp;D_Thumb_274x180_144DPI
              Fundamental Materials Research
              Novel materials are investigated at increasingly smaller scales for maximum control of their physical and chemical properties. Electron microscopy provides researchers with key insight into a wide variety of material characteristics at the micro- to nano-scale.
              Quality control and failure analysis using electron microscopy
              Quality control and failure analysis
              Quality control and assurance are essential in modern industry. We offer a range of EM and spectroscopy tools for multi-scale and multi-modal analysis of defects, allowing you to make reliable and informed decisions for process control and improvement.
              Techniques
              Large Volume Analysis
              A novel serial block-face imaging (SBFI) solution that combines multi-energy deconvolution scanning electron microscopy (MED-SEM) with in situ sectioning. Automation and ease-of-use functions provide isotropic resolution for large volume samples.
              Learn more ›
              Large Volume Analysis
              A novel serial block-face imaging (SBFI) solution that combines multi-energy deconvolution scanning electron microscopy (MED-SEM) with in situ sectioning. Automation and ease-of-use functions provide isotropic resolution for large volume samples.
              Learn more ›
              Contact us
              Style Sheet to change Applications H3 to p with em-h3-header class
              Style Sheet to change H3 to p with em-h3-header class
              Style Sheet to change H2 style to p with em-h2-header class
              Style Sheet for Komodo Tabs
              Electron microscopy services
              To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
              Learn more ›
              Style Sheet for Support and Service footer
              Style Sheet for Fonts
              Style Sheet for Cards