False-color image of free Ca2+ concentration in a Purkinje neuron from embryonic mouse cerebellum.
False-color image of free Ca2+ concentration in a Purkinje neuron from embryonic mouse cerebellum. Neurons were grown in dispersed tissue culture for 12 days, loaded with the pentapotassium salt of fura-2 (Cat. no. F1200) using a microelectrode and then challenged with trans-ACPD, an agonist of metabotropic glutamate receptors, in the absence of extracellular Ca2+. The composite image, which represents the ratio of images obtained with excitation at 340 nm and 380 nm, reveals the mobilization of internal Ca2+ stores without contribution from Ca2+ influx. The image was contributed by D.J. Linden, Johns Hopkins University, and M. Smeyne and J.A. Connor, Roche Institute of Molecular Biology.
False-color image of free Ca2+ concentration in a Purkinje neuron from embryonic mouse cerebellum. Neurons were grown in dispersed tissue culture for 12 days, loaded with the pentapotassium salt of fura-2 (Cat. no. F1200) using a microelectrode and then challenged with trans-ACPD, an agonist of metabotropic glutamate receptors, in the absence of extracellular Ca2+. The composite image, which represents the ratio of images obtained with excitation at 340 nm and 380 nm, reveals the mobilization of internal Ca2+ stores without contribution from Ca2+ influx. The image was contributed by D.J. Linden, Johns Hopkins University, and M. Smeyne and J.A. Connor, Roche Institute of Molecular Biology.
Related Products
Catalog # Name Size Price (USD) Qty
F1200 Fura-2, Pentapotassium Salt, cell impermeant Each
240.65
Online Exclusive
257.00
Save 16.35 (6%)
Related Images