Product References
Epigenetic biomarker screening by FLIM-FRET for combination therapy in ER+ breast cancer.
Clinical epigenetics
Liu W,Cui Y,Ren W,Irudayaraj J
Research topic: Breast Cancer
Wed Jan 30 00:00:00 UTC 2019
Enhanced immunological detection of epigenetic modifications of DNA in healthy and cancerous cells by fluorescence microscopy.
Microscopy research and technique
Çelik-Uzuner S
Research topic: Methods Paper
Fri Nov 01 00:00:00 UTC 2019
TET1 regulates fibroblast growth factor 8 transcription in gonadotropin releasing hormone neurons.
PloS one
Linscott ML,Chung WCJ
Research topic: Kallmann Syndrome (KS), Gene Regulation
Tue Mar 10 00:00:00 UTC 2020
The Histone Demethylase KDM5 Is Essential for Larval Growth in Drosophila.
Genetics
Drelon C,Belalcazar HM,Secombe J
Research topic: DNA Methylation
Sun Jul 01 00:00:00 UTC 2018
Loss of Fam60a, a Sin3a subunit, results in embryonic lethality and is associated with aberrant methylation at a subset of gene promoters.
eLife
Nabeshima R,Nishimura O,Maeda T,Shimizu N,Ide T,Yashiro K,Sakai Y,Meno C,Kadota M,Shiratori H,Kuraku S,Hamada H
Research topic: Cancer - Lung
Thu Aug 02 00:00:00 UTC 2018
The Cancer/Testes (CT) Antigen HORMAD1 promotes Homologous Recombinational DNA Repair and Radioresistance in Lung adenocarcinoma cells.
Scientific reports
Gao Y,Kardos J,Yang Y,Tamir TY,Mutter-Rottmayer E,Weissman B,Major MB,Kim WY,Vaziri C
Research topic: Developmental Biology
Wed Oct 17 00:00:00 UTC 2018
Decrease of 5-hydroxymethylcytosine and TET1 with nuclear exclusion of TET2 in small intestinal neuroendocrine tumors.
BMC cancer
Barazeghi E,Prabhawa S,Norlén O,Hellman P,Stålberg P,Westin G
Research topic: Small Intestinal Neuroendocrine Tumors
Wed Jul 25 00:00:00 UTC 2018
Tumour hypoxia causes DNA hypermethylation by reducing TET activity.
Nature
Thienpont B,Steinbacher J,Zhao H,D'Anna F,Kuchnio A,Ploumakis A,Ghesquière B,Van Dyck L,Boeckx B,Schoonjans L,Hermans E,Amant F,Kristensen VN,Peng Koh K,Mazzone M,Coleman M,Carell T,Carmeliet P,Lambrechts D
Research topic: Hypoxia
Thu Sep 01 00:00:00 UTC 2016
5-Hydroxymethylcytosine discriminates between parathyroid adenoma and carcinoma.
Clinical epigenetics
Barazeghi E,Gill AJ,Sidhu S,Norlén O,Dina R,Palazzo FF,Hellman P,Stålberg P,Westin G
Research topic: Parathyroid
Fri Apr 22 00:00:00 UTC 2016
5-Hydroxymethylcytosine discriminates between parathyroid adenoma and carcinoma.
Clinical epigenetics
Barazeghi E,Gill AJ,Sidhu S,Norlén O,Dina R,Palazzo FF,Hellman P,Stålberg P,Westin G
Research topic: Parathyroid
Fri Apr 22 00:00:00 UTC 2016
Tet1 and Tet2 Protect DNA Methylation Canyons against Hypermethylation.
Molecular and cellular biology
Wiehle L,Raddatz G,Musch T,Dawlaty MM,Jaenisch R,Lyko F,Breiling A
Research topic: DNA Methylation
Mon Feb 01 00:00:00 UTC 2016
Tet1 and Tet2 Protect DNA Methylation Canyons against Hypermethylation.
Molecular and cellular biology
Wiehle L,Raddatz G,Musch T,Dawlaty MM,Jaenisch R,Lyko F,Breiling A
Research topic: DNA Methylation
Mon Feb 01 00:00:00 UTC 2016
DNA hydroxymethylation controls cardiomyocyte gene expression in development and hypertrophy.
Nature communications
Greco CM,Kunderfranco P,Rubino M,Larcher V,Carullo P,Anselmo A,Kurz K,Carell T,Angius A,Latronico MV,Papait R,Condorelli G
Research topic: Cardiovascular
Thu Aug 04 00:00:00 UTC 2016
DNA hydroxymethylation controls cardiomyocyte gene expression in development and hypertrophy.
Nature communications
Greco CM,Kunderfranco P,Rubino M,Larcher V,Carullo P,Anselmo A,Kurz K,Carell T,Angius A,Latronico MV,Papait R,Condorelli G
Research topic: Cardiovascular
Thu Aug 04 00:00:00 UTC 2016
5-Carboxylcytosine levels are elevated in human breast cancers and gliomas.
Clinical epigenetics
Eleftheriou M,Pascual AJ,Wheldon LM,Perry C,Abakir A,Arora A,Johnson AD,Auer DT,Ellis IO,Madhusudan S,Ruzov A
Research topic: Breast Cancer, Glioma
Mon Aug 24 00:00:00 UTC 2015
5-Carboxylcytosine levels are elevated in human breast cancers and gliomas.
Clinical epigenetics
Eleftheriou M,Pascual AJ,Wheldon LM,Perry C,Abakir A,Arora A,Johnson AD,Auer DT,Ellis IO,Madhusudan S,Ruzov A
Research topic: Breast Cancer, Glioma
Mon Aug 24 00:00:00 UTC 2015
Structure of a Naegleria Tet-like dioxygenase in complex with 5-methylcytosine DNA.
Nature
Hashimoto H,Pais JE,Zhang X,Saleh L,Fu ZQ,Dai N,Corrêa IR,Zheng Y,Cheng X
Research topic: structural analysis of DNA/NgTet
Thu Feb 20 00:00:00 UTC 2014
TET-mediated oxidation of methylcytosine causes TDG or NEIL glycosylase dependent gene reactivation.
Nucleic acids research
Müller U,Bauer C,Siegl M,Rottach A,Leonhardt H
Research topic: Gene Regulation
Tue Jul 01 00:00:00 UTC 2014
Alteration of genic 5-hydroxymethylcytosine patterning in olfactory neurons correlates with changes in gene expression and cell identity.
Proceedings of the National Academy of Sciences of the United States of America
Colquitt BM,Allen WE,Barnea G,Lomvardas S
Research topic: Neurobiology
Tue Sep 03 00:00:00 UTC 2013