View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 2
Heat-mediated antigen retrieval using a sodium citrate buffer (pH 6.0) is recommended for the staining of paraffin sections. For FACS analysis, use 10 µL of the suggested working dilution to label 1x10^6 cells in 100 µL.
Mouse anti 5-methylcytidine antibody, clone 33D3 recognizes 5-methylcytidine, a modified base found in the DNA of plants and vertebrates.
Modified base 5-methyl cytidine (5-MeCyd) is found in DNA of plants and vertebrates. DNA methylation is a DNA modification process, which is involved in the control of gene expression. Reports suggest that in tumours, DNA is frequently globally hypomethylated compared to the DNA from normal tissue.