Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Alomone Labs, LTD
FIGURE: 1 / 2
Reconstitution: 1 X 25 µL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
In platelets and gut, serotonin plays a major role in cardiovascular function and motility of the gastrointestinal tract, respectively. Serotonin mediates its effects through several of G protein coupled receptors, designated 5-HT receptors or alternatively SR receptors. The SR-2 receptors are comprised of three subtypes, SR-2A, SR-2B and SR-2C, which activate phospholipase C and release intracellular stores of calcium in response to serotonin. SR-2A has a specific role in tracheal smooth muscle contraction, bronchoconstriction and mediating aldosterone production, and it is also thought to play a role in several psychiatric disorders, including depression and schizophrenia. SR-2B is expressed in embryonic and adult cardiovascular tissues, gut and brain and plays an important role in the pathology of cardiac disorders. SR-2C is thought to mediate the effects of atypical antipsychotic drugs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5-HT-2; 5-HT-2A; 5-HT2 receptor; 5-HT2A receptor; 5-HT2A/2C; 5-HT2A/2C receptor; 5-hydroxytryptamine (serotonin) receptor 2A, G protein-coupled; 5-hydroxytryptamine 2 receptor; 5-hydroxytryptamine receptor 2A; 5HT2A Receptor; serotonin 5-HT-2A receptor; serotonin 5HT-2 receptor; Serotonin receptor 2A; SR2A
Gene Aliases: 5-HT2A; 5Ht-2; E030013E04; Htr-2; HTR2; HTR2A
UniProt ID: (Human) P28223, (Mouse) P35363, (Rat) P14842
Entrez Gene ID: (Human) 3356, (Mouse) 15558, (Rat) 29595
Molecular Function: G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support