Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The antibody detects endogenous levels of total A26C2 + A26C3 protein.
Belongs to the POTE family. 3 isoforms of the human protein are produced by alternative splicing. Bera T.K., Gene 337:45-53(2004).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: actin, beta-like 1; ANKRD26-like family C member 2; ANKRD26-like family C member 3; ANKRD26-like family C, member 2; ANKRD26-like family C, member 3; cancer/testis antigen family 104, member 4; cancer/testis antigen family 104, member 7; POTE ankyrin domain family member G; POTE ankyrin domain family member H; POTE ankyrin domain family, member G; POTE ankyrin domain family, member H; POTE-14; POTE-22; POTE14B; Prostate, ovary, testis-expressed protein on chromosome 14; Prostate, ovary, testis-expressed protein on chromosome 22; protein expressed in prostate, ovary, testis, and placenta 14; protein expressed in prostate, ovary, testis, and placenta 22; protein expressed in prostate, ovary, testis, and placenta POTE14 like
Gene Aliases: A26C2; A26C3; ACTBL1; CT104.4; CT104.7; POTE-14; POTE14; POTE14alpha; POTE22; POTEG; POTEH
UniProt ID: (Human) Q6S5H5, (Human) Q6S545
Entrez Gene ID: (Human) 404785, (Human) 23784
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support