Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: FSPMEGTNQL LLQQPLVKKV EFGTDTLNIY LDELIKNTQT YTFTISQSVL VTNLKPATIK VYDYYLPDEQ ATIQYSDPCE
A2ML1 is a new member of the alpha 2-macroglobulin protease inhibitor family, and shares most the characteristics of this family. It is the first family member detected in the epidermis and may play an important role during desquamation by inhibiting extracellular proteases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha-2-macroglobulin-like 1; Alpha-2-macroglobulin-like protein 1; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 9; C3 and PZP-like, alpha-2-macroglobulin domain containing 9
Gene Aliases: A2ML1; CPAMD9
UniProt ID: (Human) Q2M224
Entrez Gene ID: (Human) 144568
Molecular Function: protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support