Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MRKELQLSLS VTLLLVCGFL YQFTLKSSCL FCLPSFKSHQ GLEALLSHRR GIVFLETSER MEPPHLVSCS VESAAKIYPE WPVVFFMKGL TDSTPMPSNS TYPAFSFLSA IDNVFLFPLD MKRLLEDTPL FSWYNQINAS AERNWLHISS DASRLAIIWK YGGIYMDTDV ISIRPIPEEN FLAAQASRYS SNGIFGFLPH HPFLWECMEN FVEHYNSAIW GNQGPELMTR MLRVWCKLED FQEVSDLRCL NISFLHPQRF YPISYREWRR YYEVWDTEPS FNVSYALHLW NHMNQEGRAV IRGSNTLVEN LYRKHCPRTY RDLIKGPEGS VTGELGPGNK; Positive Samples: SGC-7901, 293T, LO2; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein
This gene encodes a protein from the glycosyltransferase 32 family. The enzyme catalyzes the transfer of N-acetylglucosamine to core 2 branched O-glycans. It forms a unique glycan, GlcNAcalpha1->4Galbeta->R and is largely associated with the Golgi apparatus membrane.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-1,4-N-acetylglucosaminyltransferase; alpha4 GnT; Alpha4GnT; GnTalpha4; MGC149493
Gene Aliases: A4GNT; alpha4GnT
UniProt ID: (Human) Q9UNA3
Entrez Gene ID: (Human) 51146
Molecular Function:
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support