Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
The protein encoded by this gene belongs to the class-II aminoacyl-tRNA synthetase family. Aminoacyl-tRNA synthetases play critical roles in mRNA translation by charging tRNAs with their cognate amino acids. The encoded protein is a mitochondrial enzyme that specifically aminoacylates alanyl-tRNA. Mutations in this gene are a cause of combined oxidative phosphorylation deficiency 8. Catalyzes the attachment of alanine to tRNA(Ala) in a two-step reaction: alanine is first activated by ATP to form Ala-AMP and then transferred to the acceptor end of tRNA(Ala). Also edits incorrectly charged tRNA(Ala) via its editing domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AARS2; alanine tRNA ligase 2, mitochondrial (putative); Alanine--tRNA ligase, mitochondrial; alanine--tRNA ligase, mitochondrial {ECO:0000255|HAMAP-Rule:MF_03133}; Alanyl-tRNA synthetase; alanyl-tRNA synthetase 2, mitochondrial (putative); alanyl-tRNA synthetase like; alanyl-tRNA synthetase {ECO:0000255|HAMAP-Rule:MF_03133}; alanyl-tRNA synthetase, mitochondrial; AlaRS; alaRS {ECO:0000255|HAMAP-Rule:MF_03133}; bA444E17.1; probable alanyl-tRNA synthetase, mitochondrial
Gene Aliases: AARS2; AARSL; AlaRS; COXPD8; Gm89; KIAA1270; LKENP; MT-ALARS; MTALARS
UniProt ID: (Human) Q5JTZ9, (Mouse) Q14CH7, (Rat) D3ZX08
Entrez Gene ID: (Human) 57505, (Mouse) 224805, (Rat) 301254
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support