Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: VQSISDFEKF TKGMDDLGSS EEEEDEESGM EEGDDAEDSQ GESEEDRAGD RNSEDDGVVM TFSSVKVSEE VEKGRAVKNQ IALWDQLLEG RIKLQKALLT
The protein encoded by this gene was identified on the basis of its interaction with MAP3K12/DLK, a protein kinase known to be involved in the induction of cell apoptosis. This gene product contains a leucine zipper, which is a characteristic motif of transcription factors, and was shown to exhibit strong transactivation activity when fused to Gal4 DNA binding domain. Overexpression of this gene interfered with MAP3K12 induced apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apoptosis-antagonizing transcription factor; Protein AATF; Rb-binding protein Che-1
Gene Aliases: AATF; BFR2; CHE-1; CHE1; DED; HSPC277
UniProt ID: (Human) B3KQ26
Entrez Gene ID: (Human) 26574
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support