Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: SK-N-SH, mouse brain, rat liver.
Predicted reactivity: Mouse (93%), Rat (93%), Xenopus laevis (82%), Pig (95%), Bovine (99%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: (S)-3-amino-2-methylpropionate transaminase; 4-aminobutyrate aminotransferase, mitochondrial; 4-aminobutyrate transaminase; beta-AlaAT I; beta-alanine oxoglutarate aminotransferase; FLJ17813; FLJ30272; GABA aminotransferase; GABA AT; GABA T; GABA transaminase; GABA transferase; GABA-AT; GABA-T; Gamma-amino-N-butyrate transaminase; hCG1984265; L AIBAT; L-AIBAT
Gene Aliases: 9630038C02Rik; ABAT; AI255750; beta-AlaAT; ENSMUSG00000051226; GABA-AT; Gabaat; GABAT; Gm9851; I54; Laibat; NPD009; X61497
UniProt ID: (Human) P80404, (Rat) P50554, (Mouse) P61922
Entrez Gene ID: (Human) 18, (Rat) 81632, (Mouse) 268860
Molecular Function: transaminase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support