Product References
Targeting Cholesterol Homeostasis Improves Recovery in Experimental Optic Neuritis.
Biomolecules
Godwin CR,Anders JJ,Cheng L,Elwood BW,Kardon RH,Gramlich OW
Published figure using ABCA1 polyclonal antibody (Product # PA1-16789) in Immunohistochemistry
Fri Oct 07 00:00:00 EDT 2022
Givinostat-Liposomes: Anti-Tumor Effect on 2D and 3D Glioblastoma Models and Pharmacokinetics.
Cancers
Taiarol L,Bigogno C,Sesana S,Kravicz M,Viale F,Pozzi E,Monza L,Carozzi VA,Meregalli C,Valtorta S,Moresco RM,Koch M,Barbugian F,Russo L,Dondio G,Steinkühler C,Re F
PA1-16789 was used in Western Blotting to provide the proof of concept for a delivery system that can improve the in vivo half-life and increase the brain delivery of Givinostat, a pan-HDAC inhibitor.
Thu Jun 16 00:00:00 EDT 2022
Liver RBFOX2 regulates cholesterol homeostasis via Scarb1 alternative splicing in mice.
Nature metabolism
Paterson HAB,Yu S,Artigas N,Prado MA,Haberman N,Wang YF,Jobbins AM,Pahita E,Mokochinski J,Hall Z,Guerin M,Paulo JA,Ng SS,Villarroya F,Rashid ST,Le Goff W,Lenhard B,Cebola I,Finley D,Gygi SP,Sibley CR,Vernia S
PA1-16789 was used in ChIP Assay, ELISA, Flow Cytometry, Gel Shift, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Neutralization, Western Blot to identify a key role for the splicing factor RNA-binding Fox protein 2 (RBFOX2) in maintaining cholesterol homeostasis in a lipogenic environment in the liver.
Thu Dec 01 00:00:00 EST 2022
CTRP12 ameliorates atherosclerosis by promoting cholesterol efflux and inhibiting inflammatory response via the miR-155-5p/LXRα pathway.
Cell death & disease
Wang G,Chen JJ,Deng WY,Ren K,Yin SH,Yu XH
Published figure using ABCA1 polyclonal antibody (Product # PA1-16789) in Western Blot
Wed Mar 10 00:00:00 EST 2021
CTRP12 ameliorates atherosclerosis by promoting cholesterol efflux and inhibiting inflammatory response via the miR-155-5p/LXRα pathway.
Cell death & disease
Wang G,Chen JJ,Deng WY,Ren K,Yin SH,Yu XH
Published figure using ABCA1 polyclonal antibody (Product # PA1-16789) in Western Blot
Wed Mar 10 00:00:00 EST 2021
Liver X Receptor activation regulates genes involved in lipid homeostasis in developing chondrocytes.
Osteoarthritis and cartilage open
Sun MM,Beier F
Published figure using ABCA1 polyclonal antibody (Product # PA1-16789) in Immunohistochemistry
Sun Mar 01 00:00:00 EST 2020
Liver X Receptor activation regulates genes involved in lipid homeostasis in developing chondrocytes.
Osteoarthritis and cartilage open
Sun MM,Beier F
Published figure using ABCA1 polyclonal antibody (Product # PA1-16789) in Immunohistochemistry
Sun Mar 01 00:00:00 EST 2020
PMP22 Regulates Cholesterol Trafficking and ABCA1-Mediated Cholesterol Efflux.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Zhou Y,Miles JR,Tavori H,Lin M,Khoshbouei H,Borchelt DR,Bazick H,Landreth GE,Lee S,Fazio S,Notterpek L
PA1-16789 was used in Immunocytochemistry-immunoflourescence to reveal a novel role for PMP22 in regulating lipid metabolism and cholesterol trafficking through functional interaction with the cholesterol efflux regulatory protein, ABCA1.
Wed Jul 03 00:00:00 EDT 2019
PMP22 Regulates Cholesterol Trafficking and ABCA1-Mediated Cholesterol Efflux.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Zhou Y,Miles JR,Tavori H,Lin M,Khoshbouei H,Borchelt DR,Bazick H,Landreth GE,Lee S,Fazio S,Notterpek L
PA1-16789 was used in Immunocytochemistry-immunoflourescence to reveal a novel role for PMP22 in regulating lipid metabolism and cholesterol trafficking through functional interaction with the cholesterol efflux regulatory protein, ABCA1.
Wed Jul 03 00:00:00 EDT 2019
The Liver X Receptor Is Upregulated in Monocyte-Derived Macrophages and Modulates Inflammatory Cytokines Based on LXRα Polymorphism.
Mediators of inflammation
Kim HA,Baek WY,Han MH,Jung JY,Suh CH
PA1-16789 was used in Western Blotting to suggest that the expression levels of LXRα, according to LXRα -1830 T > C genotype, may contribute to the inflammatory response by induction of inflammatory cytokines in SLE.
Mon Aug 05 00:00:00 EDT 2019
ATP Binding Cassette Transporter A1 is Involved in Extracellular Secretion of Acetylated APE1/Ref-1.
International journal of molecular sciences
Lee YR,Joo HK,Lee EO,Cho HS,Choi S,Kim CS,Jeon BH
PA1-16789 was used in Western blotting to show acetylation of APE1/Ref-1 is considered to be necessary for its extracellular targeting via non-classical secretory pathway using the ABCA1 transporter.
Fri Jun 28 00:00:00 EDT 2019
Calpains Released by T Lymphocytes Cleave TLR2 To Control IL-17 Expression.
Journal of immunology (Baltimore, Md. : 1950)
Perez J,Dansou B,Hervé R,Levi C,Tamouza H,Vandermeersch S,Demey-Thomas E,Haymann JP,Zafrani L,Klatzmann D,Boissier MC,Letavernier E,Baud L
PA1-16789 was used in western blot to elucidate the role of calpains from T lymphocytes and IL-17 expression
Fri Jan 01 00:00:00 EST 2016
Xanthine oxidoreductase is involved in macrophage foam cell formation and atherosclerosis development.
Arteriosclerosis, thrombosis, and vascular biology
Kushiyama A,Okubo H,Sakoda H,Kikuchi T,Fujishiro M,Sato H,Kushiyama S,Iwashita M,Nishimura F,Fukushima T,Nakatsu Y,Kamata H,Kawazu S,Higashi Y,Kurihara H,Asano T
PA1-16789 was used in western blot to study the role of xanthine oxidoreductase in macrophage foam cell formation and the effect of a xanthine oxidireductase inhibitor on atherosclerosis
Wed Feb 01 00:00:00 EST 2012
Binding of PDZ-RhoGEF to ATP-binding cassette transporter A1 (ABCA1) induces cholesterol efflux through RhoA activation and prevention of transporter degradation.
The Journal of biological chemistry
Okuhira K,Fitzgerald ML,Tamehiro N,Ohoka N,Suzuki K,Sawada J,Naito M,Nishimaki-Mogami T
Published figure using ABCA1 polyclonal antibody (Product # PA1-16789) in Western Blot
Fri May 21 00:00:00 EDT 2010