Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EDIRHFNIF DSVLDLWAAC LYRSCLLLGA TIGVAKNSAL GPRRLRASWL VITLVCLFVG IYAMVKLLLF SEVRRPIRDP WFWALFVWTY ISLGASFLLW WLLSTVRPGT QALEPGAATE AEGFPGSGRP PPEQASGATL QKLLSYT (36-181 aa encoded by BC017348)
The structure specific ERCC 1 / XPF endonuclease complex is implicated in the repair of two distinct types of lesions in DNA: NER for UV induced lesions and bulky chemical adducts; and recombination repair of the very genotoxic interstrand cross links. NER mechanism involves dual incisions on both sides of the damage catalyzed by two nucleases. In mammalian cells, XPG cleaves 3' of the DNA lesion while the ERCC1 / XPF complex makes the 5' incision. ERCC1 / XPF probably plays a role in processing partially processed recombination intermediates.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ABC transporter 9 protein; ABC-type oligopeptide transporter ABCB9; ATP-binding cassette sub-family B member 9; ATP-binding cassette transporter 9; ATP-binding cassette, sub-family B (MDR/TAP), member 9; ATP-binding cassette, sub-family B, member 9; mABCB9; TAP-like protein; TAPL
Gene Aliases: ABCB9; EST122234; KIAA1520; mKIAA1520; TAPL
UniProt ID: (Human) Q9NP78, (Mouse) Q9JJ59
Entrez Gene ID: (Human) 23457, (Mouse) 56325
Molecular Function:
ATP-binding cassette (ABC) transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support