Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPKAPKQQPP EPEWIGDGES TSPSDKVVKK GKKDKKIKKT FFEELAVEDK QAGEEEKVLK EKEQQQQQQQ QQQKKKRDTR KGRRKKDVDD DGEEKELMER LKKLSVPTSD EEDEVPAPKP RGGKKTKGGN VFAALIQDQS EEEEEEEKHP PKPAKPEKNR INKAVPEEQQ PALKGKKGKE EKSKGKAKPQ NKFAALDNEE EDKEEEIIKE KEPPKQGKEK AKKAEQGSEE EGEGEEEEEE GGESKADDPY AHLSKKEKKK LKKQMEYERQ VASLKAANAA ENDFSVSQAE MSSRQAMLEN ASDIKLEKFS ISAHGKELFV NADLYIVAGR RYGLVGPNGK GKTTLLKHIA NRALSIPPNI DVLLCEQEVV ADETPAVQAV LRADTKRLKL LEEERRLQGQ LEQGDDTAAE RLEKVYEELR ATGAAAAEAK ARRILAGLGF DPEMQNRPTQ KFSGGWRMRV SLARALFMEP TLLMLDEPTN HLDLNAVIWL NNYLQGWRKT LLIVSHDQGF LDDVCTDIIH LDAQRLHYYR GNYMTFKKMY QQKQKELLKQ YEKQEKKLKE LKAGGKSTKQ AEKQTKEALT RKQQKCRRKN QDEESQEAPE LLKRPKEYTV RFTFPDPPPL SPPVLGLHGV TFGYQGQKPL FKNLDFGIDM DSRICIVGPN GVGKSTLLLL LTGKLTPTHG EMRKNHRLKI GFFNQQYAEQ LRMEETPTEY LQRGFNLPYQ DARKCLGRFG LESHAHTIQI CKLSGGQKAR VVFAELACRE PDVLILDEPT NNLDIESIDA LGEAINEYKG AVIVVSHDAR LITETNCQLW VVEEQSVSQI DGDFEDYKRE VLEALGEVMV SRPRE
This gene encodes a component of the spliceosome complex and is one of several retinitis pigmentosa-causing genes. When the gene product is added to the spliceosome complex, activation occurs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-binding cassette 50; ATP-binding cassette 50 (TNF-alpha stimulated); ATP-binding cassette sub-family F member 1; ATP-binding cassette, sub-family F (GCN20), member 1; TNF-alpha-stimulated ABC protein; TNFalpha-inducible ATP-binding protein
Gene Aliases: ABC27; ABC50; ABCF1
UniProt ID: (Human) Q8NE71
Entrez Gene ID: (Human) 23
Molecular Function: translation elongation factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support