Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATP-binding cassette (ABC) transporter genes are involved in the regulation of the amount of dietary cholesterol retained in the body. ABCG8, expressed at high levels in the liver and intestine, normally cooperates with ABCG5 to limit intestinal absorption and promote biliary excretion of sterols. The mutated form of this transporter can lead to sterol accumulation and atherosclerosis or sitosterolemia, a rare autosomal recessive disorder, characterized by hyperabsorption of sterols and the inability to excrete sterols into bile.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-binding cassette sub-family G member 8; ATP-binding cassette, sub-family G (WHITE), member 8; ATP-binding cassette, subfamily G, member 8; MGC142217; sterolin 2; Sterolin-2; Sterolin2
Gene Aliases: 1300003C16Rik; ABCG8; AI114946; GBD4; sterolin-2; STSL
UniProt ID: (Human) Q9H221, (Mouse) Q9DBM0
Entrez Gene ID: (Human) 64241, (Mouse) 67470
Molecular Function:
ATP-binding cassette (ABC) transporter
primary active transporter
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support