Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MLSSFLSPQN GTWADTFSLL LALAVALYLG YYWACVLQRP RLVAGPQFLA FLEPHCSITT ETFYPTLWCF EGRLQSIFQV LLQSQPLVLY QSDILQTPDG GQLLLDWAKQ PDSSQDPDPT TQPIVLLLPG ITGSSQETYV LHLVNQALRD GYQAVVFNNR GCRGEELRTH RAFCASNTED LETVVNHIKH RYPQAPLLAV GISFGGILVL NHLAQARQAA GLVAALTLSA CWDSFETTRS LETPLNSLLF NQPLTAGLCQ LVERNRKVIE KVVDIDFVLQ ARTIRQFDER YTSVAFGYQD CVTYYKAASP RTKIDAIRIP VLYLSAADDP FSPVCALPIQ AAQHSPYVAL LITARGGHIG FLEGLLPWQH WYMSRLLHQY AKAIFQDPEG LPDLRALLPS EDRNS
This gene is a member of the AB hydrolase superfamily and encodes a protein with an alpha/beta hydrolase fold. This domain is common to a number of hydrolytic enzymes of widely differing phylogenetic origins and catalytic functions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Abhydrolase domain-containing protein 1; Alpha/beta hydrolase domain-containing protein 1; Lung alpha/beta hydrolase 1; Protein ABHD1; testicular tissue protein Li 5
Gene Aliases: ABHD1; LABH1
UniProt ID: (Human) Q96SE0
Entrez Gene ID: (Human) 84696
Molecular Function: serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support