Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 3
Antibody detects endogenous levels of total ABHD1.
This gene is a member of the AB hydrolase superfamily and encodes a protein with an alpha/beta hydrolase fold. This domain is common to a number of hydrolytic enzymes of widely differing phylogenetic origins and catalytic functions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Abhydrolase domain-containing protein 1; alpha/beta hydrolase domain containing protein 1; Alpha/beta hydrolase domain-containing protein 1; alpha/beta hydrolase-1; Lung alpha/beta hydrolase 1; MmLABH1; Protein ABHD1; testicular tissue protein Li 5
Gene Aliases: ABHD1; LABH-1; LABH1
UniProt ID: (Human) Q96SE0, (Rat) Q5RK23, (Mouse) Q9QZC8
Entrez Gene ID: (Human) 84696, (Rat) 313917, (Mouse) 57742
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support