Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Peptide sequence: DLPGFGNSAP SKEASTEAGR AALLERALRD LEVQNAVLVS PSLSGHYALP
Sequence homology: Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 100%; Rat: 86%
ABHD14A play a possible role in granule neuron development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Abhydrolase domain-containing protein 14A; Alpha/beta hydrolase domain-containing protein 14A; Protein ABHD14A
Gene Aliases: ABHD14A; DORZ1; UNQ1913/PRO4373
UniProt ID: (Human) Q9BUJ0
Entrez Gene ID: (Human) 25864
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support