Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MGGVLIPSP GRVAAEWEVQ NRIPSGTILK ALMEGGENGP WMRFMRAEIT AEGFLREFGR LCSEMLKTSV PVDSFFSLLT SERVAKQFPV MTEAITQIRA KGLQTAVLSN NFYLPNQKSF LPLDRKQFDV IVESCMEGIC KPDPRIYKLC LEQLGLQPSE SIFLDDLGTN LKEAARLGIH TIKVNDPETA VKELEALLGF TLRVGVPNTR PVKKTMEIPK DSLQKYLKDL LGIQTTGL (1-237 aa encoded by BC015056)
This gene encodes a member of the acyl-CoA dehydrogenase family of enzymes (ACADs), which participate in the beta-oxidation of fatty acids in mitochondria. The encoded enzyme contains a hydrolase domain at the N-terminal portion, a serine/threonine protein kinase catalytic domain in the central region, and a conserved ACAD domain at the C-terminus. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACAD-10; Acyl-CoA dehydrogenase family member 10; acyl-CoA dehydrogenase family, member 10; acyl-Coenzyme A dehydrogenase family, member 10
Gene Aliases: 2410021P16Rik; ACAD10; RGD1310159
UniProt ID: (Human) Q6JQN1, (Mouse) Q8K370
Entrez Gene ID: (Human) 80724, (Mouse) 71985, (Rat) 304500
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support