Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: VDASVAVFC EIQNTLINTL IRKHGTEEQK ATYLPQLTTE KVGSFCLSEA GAGSDSFALK TRADKEGDYY VLNGSKMWIS SAEHAGLFLV MANVDPTIGY KGITSFLVDR DTPGLHIGKP ENKLGLRASS TCPLTFENVK VPEANILGQI GHGYKYAIGS LNEGRIGIAA QMLGLAQGCF DYTIPYIKER IQFGKRLFDF QGLQHQVAHV ATQLEAARLL TYNAARLLEA GKPFIKEASM AKYYASEIAG QTTSKCIEWM GGVGYTKDYP VEKYFRDAKI GTIYEGASNI QLNTIAKHID AEY (131-432 aa encoded by BC013756)
Short/branched chain acyl-CoA dehydrogenase(ACADSB) is a member of the acyl-CoA dehydrogenase family of enzymes that catalyze the dehydrogenation of acyl-CoA derivatives in the metabolism of fatty acids or branch chained amino acids. Substrate specificity is the primary characteristic used to define members of this gene family. The ACADSB gene product has the greatest activity towards the short branched chain acyl-CoA derivative, (S)-2-methylbutyryl-CoA, but also reacts significantly with other 2-methyl branched chain substrates and with short straight chain acyl-CoAs. The cDNA encodes for a mitochondrial precursor protein which is cleaved upon mitochondrial import and predicted to yield a mature peptide of approximately 43.7- kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-MEBCAD; 2-methyl branched chain acyl-CoA dehydrogenase; 2-methylbutyryl-CoA dehydrogenase; 2-methylbutyryl-coenzyme A dehydrogenase; acyl-CoA dehydrogenase short/branched chain; Acyl-Coenzyme A dehydrogenase short-branched chain; acyl-Coenzyme A dehydrogenase, short/branched chain; SBCAD; Short/branched chain specific acyl-CoA dehydrogenase, mitochondrial
Gene Aliases: 1300003O09Rik; 2-MEBCAD; ACAD7; ACADSB; BB066609; SBCAD
UniProt ID: (Human) P45954, (Rat) P70584, (Mouse) Q9DBL1
Entrez Gene ID: (Human) 36, (Rat) 25618, (Mouse) 66885
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support