Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive test controls include: HeLa, SW620, HepG2, Mouse liver, Mouse brain, Mouse kidney, Rat brain, Rat kidney. The target is usually found in the following locations: Cytoplasm.
Immunogen sequence: GLTDAFHNCH MGITAENVAK KWQVSREDQD KVAVLSQNRT ENAQKAGHFD KEIVPVLVST RKGLIEVKTD EFPRHGSNIE AMSKLKPYFL TDGTGTVTPA N
This gene represents a member of the 'frizzled' gene family, which encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The protein encoded by this family member contains a signal peptide, a cysteine-rich domain in the N-terminal extracellular region, and seven transmembrane domains, but unlike other family members, this protein does not contain a C-terminal PDZ domain-binding motif. This protein functions as a negative regulator of the canonical Wnt/beta-catenin signaling cascade, thereby inhibiting the processes that trigger oncogenic transformation, cell proliferation, and inhibition of apoptosis. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acetoacetyl Coenzyme A thiolase; Acetyl-CoA acetyltransferase, cytosolic; Acetyl-CoA transferase-like protein; acetyl-Coenzyme A acetyltransferase 2; acetyl-Coenzyme A acetyltransferase 3; AE1; BND3; Cytosolic acetoacetyl-CoA thiolase; DI; EMPB3; EPB3; RTA1A; SW; t-complex protein 1, related sequence 1; WD; WD1; WR
Gene Aliases: Ab2-076; ACAT2; Acat3; ACTL; AW742799; Tcp-1x; Tcp1-rs1
UniProt ID: (Human) Q9BWD1, (Rat) Q5XI22, (Mouse) Q8CAY6
Entrez Gene ID: (Human) 39, (Rat) 308100, (Mouse) 110460
Molecular Function: acyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support