Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: DLGNVLTST PNAKTVNGKA ESSDSGAESE EEEAQEEVKG AEQSDNDKKM MKKSADHKNL EVIVTNGYDK DGFVQDIQND IHASSSLNGR STEEVKPIDE NLGQTGKSAV CIHQDINDDH VEDVTGIQHL TSDSDSEVYC DSMEQFGQEE SLDSFTSNNG PFQYYLGGHS SQPMENSGFR EDIQVPPGNG NIGNMQVVAV EGKGEVKHGG EDGRNNSGAP HREKRGGETD EFSNVRRGRG HRMQHLSEGT KGRQVGSGGD GERWGSDRGS RGSLNEQIAL VLMRLQEDMQ NVLQRLQKLE TLTALQAKSS TSTLQTAPQP TSQRPSWWPF EMSP (129-461 aa encoded by B C030555)
Acyl-CoA binding protein which acts as the peroxisome receptor for pexophagy but is dispensable for aggrephagy and nonselective autophagy. Binds medium- and long-chain acyl-CoA esters.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acyl-CoA-binding domain-containing protein 5; acyl-Coenzyme A binding domain containing 5; endozepine-related protein; membrane-associated diazepam binding inhibitor
Gene Aliases: 1300014E15Rik; ACBD5; KIAA1996
UniProt ID: (Human) Q5T8D3, (Mouse) Q5XG73
Entrez Gene ID: (Human) 91452, (Mouse) 74159
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support