Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: 293T, A431, Jurkat.
Predicted reactivity: Mouse (88%), Rat (88%), Bovine (91%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the acyl-CoA thioesterase family which catalyse the conversion of activated fatty acids to the corresponding non-esterified fatty acid and coenzyme A. Expression of a mouse homolog in brown adipose tissue is induced by low temperatures and repressed by warm temperatures. Higher levels of expression of the mouse homolog has been found in obesity-resistant mice compared with obesity-prone mice, suggesting a role of acyl-CoA thioesterase 11 in obesity. Alternative splicing results in transcript variants.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acyl-CoA thioester hydrolase 11; Acyl-CoA thioesterase 11; Acyl-coenzyme A thioesterase 11; Adipose-associated thioesterase; BFIT; Brown fat-inducible thioesterase; Palmitoyl-coenzyme A thioesterase; StAR-related lipid transfer (START) domain containing 14; START domain containing 14; thioeste; thioesterase superfamily member 1; thioesterase, adipose associated
Gene Aliases: ACOT11; BFIT; KIAA0707; STARD14; THEA; THEM1
UniProt ID: (Human) Q8WXI4
Entrez Gene ID: (Human) 26027
Molecular Function: esterase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support