Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Immunogen sequence: TVLINACVA NTVAPLHYKD MIIPKLVDDL GKVKITKSGF LTFMDTWSNP LEEHNHQSLV PLEKAQVPFL FIVGMDDQSW KSEFYAQIAS E (37-126 aa encoded by BC126380)
ACOT6 gene ontology annotations related to this gene include acyl-CoA metabolic process; fatty acid metabolic process.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acyl-CoA thioesterase 6; Acyl-coenzyme A thioesterase 6; putative acyl-CoA thioesterase 6
Gene Aliases: 4632408A20Rik; A330054B07; ACOT6; BE688602; c14_5530; C14orf42; RGD1309669
UniProt ID: (Human) Q3I5F7, (Mouse) Q32Q92
Entrez Gene ID: (Human) 641372, (Rat) 299193, (Mouse) 217700
Molecular Function:
esterase
hydrolase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support