Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody detects endogenous levels of total ACOT8 protein.
Acyl-CoA thioesterases are a group of enzymes that catalyze the hydrolysis of acyl-CoAs to the free fatty acid and coenzyme A (CoASH), providing the potential to regulate intracellular levels of acyl-CoAs, free fatty acids and CoASH. It may mediate Nef-induced down-regulation of CD4. It may be involved in the metabolic regulation of peroxisome proliferation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 4,8-dimethylnonanoyl-CoA thioesterase; 48-dimethylnonanoyl-CoA thioesterase; Acyl-CoA thioesterase 8; Acyl-coenzyme A thioesterase 8; choloyl-CoA hydrolase; Choloyl-coenzyme A thioesterase; hACTE-III; HIV-Nef associated acyl-CoA thioesterase; HIV-Nef-associated acyl-CoA thioesterase; long-chain fatty-acyl-CoA hydrolase; Nef (lentivirus myristoylated factor) associated protein 1; palmitoyl-CoA hydrolase; peroxisomal acyl-CoA thioesterase 1; Peroxisomal acyl-CoA thioesterase 2; Peroxisomal acyl-coenzyme A thioester hydrolase 1; Peroxisomal long-chain acyl-CoA thioesterase 1; PTE-1; PTE-2; Thioesterase II; thioesterase III
Gene Aliases: ACOT8; ACTEIII; hACTE-III; HNAACTE; hTE; NAP1; PTE-1; PTE-2; PTE1; PTE2
UniProt ID: (Human) O14734, (Mouse) P58137
Entrez Gene ID: (Human) 10005, (Mouse) 170789, (Rat) 170588
Molecular Function: esterase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support