Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MAEQATKSVL FVCLGNICRS PIAEAVFRKL VTDQNISENW VIDSGAVSDW NVGRSPDPRA VSCLRNHGIH TAHKARQITK EDFATFDYIL CMDESNLRDL NRKSNQVKTC KAKIELLGSY DPQKQLIIED PYYGNDSDFE TVYQQCVRCC RAFLEKAH
Low molecular weight protein tyrosine phosphtases, including the human red cell phosphatase, HCPTP, are widely expressed cytosolic proteins of approximately 18 kDa that exist in distinct isoforms and are highly selective for phosphotyrosine over phosphoserine or phosphotyrosine. The HCPTP-A and HCPTP-B proteins expressed in human red cells and placenta are fast and slow forms of red cell acid phosphatase5. Like other tyrosine phosphatases, HCPTP is sensitive to inhibition by vanadate, and has a catalytic mechanism that involves formation of a cystinylphosphate intermediate6. The crystal structure of the bovine heart derived enzyme BHPTP shows a four-stranded central parallel b sheet with flanking a helices and an active site cysteine residue in the typical tyrosine phosphatase sequence context, CXXXXXR.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acid phosphatase of erythrocyte; Adipocyte acid phosphatase; cytoplasmic phosphotyrosyl protein phosphatase; HAAP betaÊ; LMW-PTP; LMW-PTPase; Low molecular weight cytosolic acid phosphatase; Low molecular weight phosphotyrosine protein phosphatase; MGC111030; MGC3499; protein tyrosine phosphatase; Red cell acid phosphatase 1; testicular secretory protein Li 37
Gene Aliases: ACP1; HAAP; LMW-PTP
UniProt ID: (Human) P24666
Entrez Gene ID: (Human) 52
Molecular Function: protein phosphatase protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support