Invitrogen antibodies—maximize lab budgets with advanced validation & coverage

Shop antibodies

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • ACSL1 Antibodies

            Invitrogen

            ACSL1 Polyclonal Antibody

            Advanced Verification
            View all (16) ACSL1 antibodies

            Promotions

            View available promotion(s)

            Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more

            Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more

            Datasheet
            Protocols
            Questions & Answers
            Catalog # PA5-78713
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            445.00
            100 µg
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite ACSL1 Polyclonal Antibody

            • Antibody Testing Data (4)
            • Advanced Verification (2)
            Application
            ACSL1 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 6

            ACSL1 Antibody (PA5-78713) in ICC/IF

            Immunofluorescence analysis of ACSL1 was performed using 70% confluent log phase MCF-10A and MCF7 cells. The cells were fixed with 4% Paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 2% BSA for 10 minutes at room temperature. The cells were labeled with ACSL1 Polyclonal Antibody (Product # PA5-78713) at 1:100 dilution in 0.1% BSA, incubated at 4 degree celsius overnight and then labeled with Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor Plus 488 (Prod... View More
            View Product

            ACSL1 Antibody in Immunocytochemistry (ICC/IF)
            ACSL1 Antibody in Western Blot (WB)
            ACSL1 Antibody in Flow Cytometry (Flow)
            ACSL1 Antibody in Flow Cytometry (Flow)
            ACSL1 Antibody
            ACSL1 Antibody

            Product Details

            PA5-78713

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            0.1-0.5 µg/mL
            -

            Immunohistochemistry (Paraffin) (IHC (P))

            0.5-1 µg/mL
            -

            Immunocytochemistry (ICC/IF)

            5 µg/mL
            -

            Flow Cytometry (Flow)

            1-3 µg/1x10^6 cells
            -
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            E. coli-derived human ACSL1 recombinant protein (Position: D604-V698).
            View immunogen

            Conjugate

            Unconjugated

            Form

            Lyophilized

            Concentration

            500 µg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS with 5mg BSA

            Contains

            0.05mg sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2745829

            Product Specific Information

            Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.

            Positive Control - WB: human HepG2 whole cell, human U87 whole cell, rat liver tissue, rat heart tissue, mouse liver tissue. IHC: mouse kidney tissue, rat kidney tissue, human liver cancer tissue. ICC/IF: U20S cell. Flow: A431 cell.

            Target Information

            The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: ACS1; Acyl CoA synthetase, long chain; Acyl-CoA synthetase 1; Arachidonate--CoA ligase; fatty acid Coenzyme A ligase, long chain 2; fatty-acid-Coenzyme A ligase, long-chain 1; fatty-acid-Coenzyme A ligase, long-chain 2; L LACS2; LACS 1; LACS 2; lignoceroyl-CoA synthase; Long-chain acyl-CoA synthetase 1; Long-chain acyl-CoA synthetase 2; Long-chain fatty acid-CoA ligase 2; long-chain fatty-acid-coenzyme A ligase 1; Long-chain-fatty-acid--CoA ligase 1; Long-chain-fatty-acid--CoA ligase, liver isozyme; OTTHUMP00000218457; OTTHUMP00000218460; OTTHUMP00000218462; OTTHUMP00000218465; Palmitoyl-CoA ligase 1; Palmitoyl-CoA ligase 2; paltimoyl-CoA ligase 1; Phytanate--CoA ligase

            View more View less

            Gene Aliases: Acas; Acas1; ACS; ACS1; ACSL1; Acsl2; COAA; FACL1; FACL2; FACS; LACS; LACS 1; LACS1; LACS2

            View more View less

            UniProt ID: (Human) P33121, (Mouse) P41216, (Rat) P18163

            View more View less

            Entrez Gene ID: (Human) 2180, (Mouse) 14081, (Rat) 25288

            View more View less

            Function(s)

            long-chain fatty acid-CoA ligase activity ATP binding decanoate--CoA ligase activity acetate-CoA ligase activity ligase

            Process(es)

            long-chain fatty acid metabolic process triglyceride metabolic process response to nutrient lipid biosynthetic process response to organic cyclic compound fatty acid transport adiponectin-activated signaling pathway response to oleic acid long-chain fatty-acyl-CoA biosynthetic process alpha-linolenic acid metabolic process xenobiotic catabolic process response to drug linoleic acid metabolic process long-chain fatty acid import positive regulation of protein serine/threonine kinase activity lipid metabolic process fatty acid metabolic process response to organic substance
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mgmkt:80/100.66.75.22:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0