Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • ACSL5 Antibodies

            Invitrogen

            ACSL5 Polyclonal Antibody

            Advanced Verification
            View all (18) ACSL5 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite ACSL5 Polyclonal Antibody

            • Antibody Testing Data (6)
            • Advanced Verification (1)
            Application
            ACSL5 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 7

            ACSL5 Antibody (PA5-52392) in ICC/IF

            Immunofluorecent analysis of ACSL5 in human cell line U-251 MG using ACSL5 Polyclonal Antibody (Product # PA5-52392). Staining shows localization to nucleoli and mitochondria.
            View Product

            ACSL5 Antibody in Immunocytochemistry (ICC/IF)
            ACSL5 Antibody in Immunohistochemistry (IHC)
            ACSL5 Antibody in Immunohistochemistry (IHC)
            ACSL5 Antibody in Immunohistochemistry (IHC)
            ACSL5 Antibody in Western Blot (WB)
            ACSL5 Antibody in Western Blot (WB)
            ACSL5 Antibody

            Product Details

            PA5-52392

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            0.04-0.4 µg/mL
            -

            Immunohistochemistry (Paraffin) (IHC (P))

            1:20-1:50
            -

            Immunocytochemistry (ICC/IF)

            0.25-2 µg/mL
            -
            Product Specifications

            Species Reactivity

            Human

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Recombinant protein corresponding to Human ACSL5. Recombinant protein control fragment (Product #RP-89597).

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.1 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.2, with 40% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2637624

            Product Specific Information

            Immunogen sequence: VHIVNKADIA MVICDTPQKA LVLIGNVEKG FTPSLKVIIL MDPFDDDLKQ RGEKSGIEIL SLYDAENLGK EHFRKPVPPS PEDLSVICFT SGTTGDPK

            Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 77%.

            Target Information

            The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. This isozyme is highly expressed in uterus and spleen, and in trace amounts in normal brain, but has markedly increased levels in malignant gliomas. This gene functions in mediating fatty acid-induced glioma cell growth. Three transcript variants encoding two different isoforms have been found for this gene.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: Arachidonate--CoA ligase; FACL5 for fatty acid coenzyme A ligase 5; fatty acid coenzyme A ligase 5; fatty-acid-Coenzyme A ligase, long-chain 5; HGNC:16526; LACS 5; Long-chain acyl-CoA synthetase 5; long-chain fatty acid coenzyme A ligase 5; Long-chain-fatty-acid--CoA ligase 5

            View more View less

            Gene Aliases: ACS2; ACS5; ACSL5; FACL5; UNQ633/PRO1250

            View more View less

            UniProt ID: (Human) Q9ULC5

            View more View less

            Entrez Gene ID: (Human) 51703

            View more View less

            Function(s)

            long-chain fatty acid-CoA ligase activity ATP binding decanoate--CoA ligase activity immunoglobulin ligase

            Process(es)

            long-chain fatty acid metabolic process long-chain fatty-acyl-CoA biosynthetic process
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-584499df9d-lm9jw:80/100.66.76.199:80.
            git-commit: 256cec658627a60c8e1196038bf5905a13cd45ba
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.33.0-2025.07.30-1.0