Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button
            Target, gene symbol, antigen
            Application
            Target Species
            • Order Status
            • Quick Order
            • Promos
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • ACSS3 Antibodies

            Invitrogen

            ACSS3 Polyclonal Antibody

            2 References
            View all (6) ACSS3 antibodies

            Promotions

            View available promotion(s)

            Promo Code: P6071074 Ending soon: buy 3, only pay for 2 Stretch your budget even further–get 33% off your purchase when you buy 3 qualifying items now through December 31. Learn more

            Promo Code: RGLOW25 Save up to $900 and get the limited-edition holiday sweatshirt + matching beanie It’s been a year – this is your badge of honor for fluorescing through it all. Offer available until December 27.. Learn more

            Spend Now, Save Big Later! Spend Now, Save Big Later! Make your New year's resolution to save: earn up to $3,500 off a future January offer when you purchase a qualifying amount before December 31st. Learn more

            Datasheet
            Protocols
            Questions & Answers
            Catalog # PA5-61631
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            373.50
            Special offer
            Online exclusive
            Ends: 13-Dec-2025
            498.00
            Save 124.50 (25%)
            100 µL
             
            Estimated availability date: 17-Dec-2025
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite ACSS3 Polyclonal Antibody

            • Antibody Testing Data (5)
            Application
            ACSS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 5

            ACSS3 Antibody (PA5-61631) in IHC (P)

            Immunohistochemical analysis of ACSS3 in human tonsil using ACSS3 Polyclonal Antibody (Product # PA5-61631) shows no positivity in non-germinal center cells as expected.
            View Product

            ACSS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            ACSS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            ACSS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            ACSS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            ACSS3 Antibody in Western Blot (WB)
            ACSS3 Polyclonal Antibody

            Product Details

            PA5-61631

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            0.04-0.4 µg/mL
            View 1 publication 1 publication

            Immunohistochemistry (Paraffin) (IHC (P))

            1:200-1:500
            -

            Immunohistochemistry (PFA fixed) (IHC (PFA))

            -
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Human

            Published species

            Mouse

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Recombinant protein corresponding to Human ACSS3. Recombinant protein control fragment (Product #RP-104026).
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.2 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.2, with 40% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2637636

            Product Specific Information

            Immunogen sequence: ASKELSSRID HVKPKVVVTA SFGIEPGRRV EYVPLVEEAL KIGQHKPDKI LIYNRPNMEA VPLAPGRDLD WDEEMAKAQS HDCVPVLSEH PLYIL

            Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 88%.

            Target Information

            Catalyzes the synthesis of acetyl-CoA from short-chain fatty acids (PubMed:28003429). Propionate is the preferred substrate (PubMed:28003429). Can utilize acetate and butyrate with a much lower affinity. [UniProt]

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: Acetate--CoA ligase 3; Acyl-CoA synthetase short-chain family member 3; Acyl-CoA synthetase short-chain family member 3, mitochondrial; AMP-binding enzyme, 33217; Propionate--CoA ligase

            View more View less

            Gene Aliases: ACSS3

            View more View less

            UniProt ID: (Human) Q9H6R3

            View more View less

            Entrez Gene ID: (Human) 79611

            View more View less

            Function(s)

            acetate-CoA ligase activity ATP binding ligase

            Process(es)

            metabolic process
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us About Us
            • Careers Careers
            • Investors Investors
            • News News
            • Social Responsibility Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
            • Policies and Notices
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-blue-555ccb4fbd-wx6js:80/100.66.76.49:80.
            git-commit: 75f0c7d4cb44686da2af39b21f2e6446a20282e2
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.41.0-2025.12.99-1.0