Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MVVERDDGST LMEIDGDKGK QGGPTYYIDT NALRVPRENM EAISPLKNGM VEDWDSFQAI LDHTYKMHVK SEASLHPVLM SEAPWNTRAK REKLTELMFE HYNIPAFFLC KTAVLTAFAN GRSTGLILDS GATHTTAIPV HDGYVLQQGI VKSPLAGDFI TMQCRELFQE MNIELVPPYM IASKEAVREG SPANWKRKEK LPQVTRSWHN YMCNCVIQDF QASVLQVSDS TYDEQVAAQM PTVHYEFPNG YNCDFGAERL KIPEGLFDPS NVKGLSGNTM LGVSHVVTTS VGMCDIDIRP GLYGSVIVAG GNTLIQSFTD RLNRELSQKT PPSMRLKLIA NNTTVERRFS SWIGGSILAS LGTFQQMWIS KQEYEEGGKQ CVERKCP
BAF53A is a family member of actin-related proteins (ARPs), which share significant amino acid sequence identity to conventional actins. Both actins and ARPs have an actin fold, which is an ATP-binding cleft, as a common feature. The ARPs are involved in diverse cellular processes, including vesicular transport, spindle orientation, nuclear migration and chromatin remodeling. This gene encodes a 53 kDa subunit protein of the BAF (BRG1/brm-associated factor) complex in mammals, which is functionally related to SWI/SNF complex in S. cerevisiae and Drosophila; the latter is thought to facilitate transcriptional activation of specific genes by antagonizing chromatin-mediated transcriptional repression. Together with beta-actin, it is required for maximal ATPase activity of BRG1, and for the association of the BAF complex with chromatin/matrix. Three transcript variants that encode two different protein isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 53 kDa BRG1-associated factor A; actin-like 6A; actin-like 6B; Actin-like protein 6A; actin-related protein 4; Actin-related protein Baf53a; ACTL6B; ArpNbeta; BAF complex 53 kDa subunit; BAF53; BAF53A; BRG1-associated factor 53A; hArpN alpha; hArpN beta; INO80 complex subunit K; MGC5382; MGC5382 Arp4
Gene Aliases: ACTL6; ACTL6A; Arp4; ARPN-BETA; BAF53; BAF53A; INO80K
UniProt ID: (Human) O96019
Entrez Gene ID: (Human) 86
Molecular Function: actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support