Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MWAPPAAIMG DGPTKKVGNQ APLQTQALQT ASLRDGPAKR AVWVRHTSSE PQEPTESKAA KERPKQEVTK AVVVDLGTGY CKCGFAGLPR PTHKISTTVG KPYMETAKTG DNRKETFVGQ ELNNTNVHLK LVNPLRHGII VDWDTVQDIW EYLFRQEMKI APEEHAVLVS DPPLSPHTNR EKYAEMLFEA FNTPAMHIAY QSRLSMYSYG RTSGLVVEVG HGVSYVVPIY EGYPLPSITG RLDYAGSDLT AYLLGLLNSA GNEFTQDQMG IVEDIKKKCC FVALDPIEEK KVPLSEHTIR YVLPDGKEIQ LCQERFLCSE MFFKPSLIKS MQLGLHTQTV SCLNKCDIAL KRDLMGNILL CGGSTMLSGF PNRLQKELSS MCPNDTPQVN VLPERDSAVW TGGSILASLQ GFQPLWVHRF EYEEHGPFFL YRRCF
The protein encoded by this gene is a member of a family of actin-related proteins (ARPs) which share significant amino acid sequence identity to conventional actins. Both actins and ARPs have an actin fold, which is an ATP-binding cleft, as a common feature. The ARPs are involved in diverse cellular processes, including vesicular transport, spindle orientation, nuclear migration and chromatin remodeling. This gene (ACTL7A), and related gene, ACTL7B, are intronless, and are located approximately 4 kb apart in a head-to-head orientation within the familial dysautonomia candidate region on 9q31. Based on mutational analysis of the ACTL7A gene in patients with this disorder, it was concluded that it is unlikely to be involved in the pathogenesis of dysautonomia. The ACTL7A gene is expressed in a wide variety of adult tissues, however, its exact function is not known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: actin-like 7-alpha; actin-like 7A; Actin-like protein 7A; Actin-like-7-alpha; testicular secretory protein Li 3
Gene Aliases: ACTL7A
UniProt ID: (Human) Q9Y615
Entrez Gene ID: (Human) 10881
Molecular Function: actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support