Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MESYDIIANQ PVVIDNGSGV IKAGFAGDQI PKYCFPNYVG RPKHMRVMAG ALEGDLFIGP KAEEHRGLLT IRYPMEHGVV RDWNDMERIW QYVYSKDQLQ TFSEEHPVLL TEAPLNPSKN REKAAEVFFE TFNVPALFIS MQAVLSLYAT GRTTGVVLDS GDGVTHAVPI YEGFAMPHSI MRVDIAGRDV SRYLRLLLRK EGVDFHTSAE FEVVRTIKER ACYLSINPQK DEALETEKVQ YTLPDGSTLD VGPARFRAPE LLFQPDLVGD ESEGLHEVVA FAIHKSDMDL RRTLFANIVL SGGSTLFKGF GDRLLSEVKK LAPKDIKIKI SAPQERLYST WIGGSILASL DTFKKMWVSK KEYEEDGSRA IHRKTF
This gene encodes a 42.3 kD subunit of dynactin, a macromolecular complex consisting of 10 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein and is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit, like ACTR1A, is an actin-related protein. These two proteins, which are of equal length and share 90% amino acid identity, are present in a constant ratio of approximately 1:15 in the dynactin complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Actin-related protein 1B; ARP1B; Beta-centractin; centractin beta; yeast homolog B
Gene Aliases: ACTR1B; ARP1B; CTRN2; PC3
UniProt ID: (Human) P42025
Entrez Gene ID: (Human) 10120
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support