Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: DKGLVDDIKK KLCYVALEPE KELSRRPEEV LREYKLPDGN IISLGDPLHQ APEALFVPQQ LGSQSPGLSN MVSSSITKCD TDIQKILFGE I
The protein encoded by this intronless gene belongs to the actin family. Studies have shown that this protein may be involved in cytoskeletal organization similar to other cytoplasmic actin-related protein (ARP) subfamily members. Antibody raised against the human protein has been used to detect the protein by immunoblotting and immunofluorescence microscopy, demonstrating its specific synthesis in the testis, late in spermatid differentiation, and its localization in the calyx.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: actin-related protein hArpM2; Actin-related protein M2; Actin-related protein T2; ARP-T2
Gene Aliases: ACTRT2; Arp-T2; ARPM2; ARPT2; HARPM2
UniProt ID: (Human) Q8TDY3
Entrez Gene ID: (Human) 140625
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support