Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTRALCSALR QALLLLAAAA ELSPGLKCVC LLCDSSNFTC QTEGACWASV MLTNGKEQVI KSCVSLPELN AQVFCHSSNN VTKTECCFTD FCNNITLHLP TASPNAPKLG PMELAIIITV PVCLLSIAAM LTVWACQGRQ CSYRKKKRPN VEEPLSECNL VNAGKTLKDL IYDVTASGSG SGLPLLVQRT IARTIVLQEI VGKGRFGEVW HGRWCGEDVA VKIFSSRDER YWFREAEIYQ TVMLRHENIL GFIAADNKDN GTWTQLWLVS EYHEQGSLYD YLNRNIVTMA GMIKLALSIA SGLAHLHMEI VGTQGKPAIA HRDIKSKNIL VKKCETCAIA DLGLAVKHDS ILNTIDIPQN PKVGTKRYMA PEMLDDTMNV NIFESFKRAD IYSVGLVYWE IARRCSVGGI VEEYQLPYYD MVPSDPSIEE MRKVVCDQKF RPSIPNQWQS CEALRVMGRI MRECWYANGA ARLTALRIKK TISQLCVKED CKA
ACVR1C, a serine/threonine protein kinase, is a type I receptor for the TGFB family of signaling molecules. The receptor complex consisting of 2 type II and 2 type I transmembrane serine/threonine kinases. Type II receptors phosphorylate and activate type I receptors which autophosphorylate, then bind and activate SMAD transcriptional regulators, SMAD2 and SMAD3, which then translocate to the nucleus and interact directly with DNA or in complex with other transcription factors. ACVR1C is a receptor for activin AB, activin B and NODAL. This protein plays a role in cell differentiation, growth arrest and apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: activin A receptor type IC; activin A receptor, type IC; Activin receptor type IC; Activin receptor type-1C; Activin receptor-like kinase 7; ACTR-IC; ACTRI; ACVR1A; ACVR1C; ACVRLK2; ALK-7; ALK2; FOP; TSRI
Gene Aliases: ACVR1C; ACVRLK7; ALK7
UniProt ID: (Human) Q8NER5
Entrez Gene ID: (Human) 130399
Molecular Function: serine/threonine protein kinase receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support