Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: HVRRRQEKQ RGLHSELGES SLILKASEQG DSMLGDLLDS DCTTGSGSGL PFLVQRTVAR QVALVECVGK GRYGEVWRGL WHGESVAVKI FSSRDEQSWF RETEIYNTVL LRHDNILGFI ASDMTSRNSS TQLWLITHYH EHGSLYDFLQ RQTLEPHLAL RLAVSAACGL AHLHVEIFGT QGKPAIAHRD FKSRNVLVKS NLQCCIADLG LAVMHSQGSD YLDIGNNPRV GTKRYMAPEV LDEQIRTDCF ESYKWTDIWA FGLVLWEIAR RTIVNGIVED YRPPFYDVVP NDPSFEDMKK VVCVDQQTPT IPNRLAADPV LSGLAQMMRE CWYPNPSARL TALRIKKTLQ KISNSPEKPK VIQ (142-503 aa encoded by BC042637)
Activin A receptor type II-like 1 (ACVRL1) is a type I cell-surface receptor for the TGF-beta superfamily of ligands. ACVRL1, similar to other type 1 receptors, has a conserved serine-threonine kinase subdomain, a glycine- and serine-rich region (called the GS domain) preceding the kinase domain, and a short C-terminal tail. Mutations in ACVRL1 are associated with hemorrhagic telangiectasia type 2. Mutations in ACVRL1 have been linked to Fibrodysplasia Ossificans Progressiva (FOP), a rare genetic disorder characterized by the formation of bone in muscles, tendons, and other connective tissues, as well as Epicanthus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: activin A receptor type II-like 1; activin A receptor type IL; activin A receptor, type II-like kinase 1; Activin receptor-like kinase 1; ALK-1; Serine/threonine-protein kinase receptor R3; SKR3; TGF-B superfamily receptor type I; TSR-I
Gene Aliases: ACVRL1; ACVRLK1; ALK-1; ALK1; HHT; HHT2; ORW2; SKR3; TSR-I
UniProt ID: (Human) P37023
Entrez Gene ID: (Human) 94
Molecular Function:
serine/threonine protein kinase receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support